Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
SFRS18 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: argi; Arginine/serine-rich protein PNISR; PNN-interacting serine/arginine-rich protein; RP11-98I9.2; serine-arginine-rich-splicing regulatory protein 130; Serine/arginine-rich-splicing regulatory protein 130; Splicing factor, arginine/serine-rich 130; Splicing factor, arginine/serine-rich 18; SR rich protein; SR-related protein; SR-rich protein; SRrp130
Gene Aliases: bA98I9.2; C6orf111; HSPC261; HSPC306; PNISR; SFRS18; SRRP130
UniProt ID: (Human) Q8TF01
Entrez Gene ID: (Human) 25957
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support