Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: RKQSNYEDLE AQPRGEEAQL SGDQLPFVME ELPGEGGNGR SEGGEAAGGP AQESRQEVRG SPRGVPLVAG SSEEGSRRSL KDAYLEVWRL VRGTRYM
Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 71%.
May play a role in hemostasis as a regulator of the biosynthesis of platelet-dense granules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frc, fringe-like 1; Fringe connection-like protein 1; Solute carrier family 35 member D3; solute carrier family 35, member D3
Gene Aliases: FRCL1; SLC35D3
UniProt ID: (Human) Q5M8T2
Entrez Gene ID: (Human) 340146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support