Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 11
SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear ″speckles″, in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delayed-early protein HRS; epididymis secretory protein Li 91; Insulin-induced growth response protein CL-4; pre-mRNA splicing factor SRp20; Pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; Pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; Serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; Serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; splicing factor arginine/serine-rich 5 (SRp40 HRS); Splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); Splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); Splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); Splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB
Gene Aliases: 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; HEL-S-91; HRS; MNCb-2616; RGD1561347; SFRS3; SFRS4; SFRS5; SFRS6; SRP20; SRP40; SRP55; SRP75; SRSF3; SRSF4; SRSF5; SRSF6; X16
UniProt ID: (Human) P84103, (Human) Q08170, (Human) Q13243, (Human) Q13247, (Mouse) P84104, (Rat) Q09167, (Mouse) O35326, (Rat) G3V6S8, (Mouse) Q3TWW8
Entrez Gene ID: (Human) 6428, (Human) 6429, (Human) 6430, (Human) 6431, (Rat) 361814, (Mouse) 20383, (Rat) 362612, (Mouse) 57317, (Rat) 29667, (Mouse) 20384, (Rat) 362264, (Mouse) 67996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support