Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Positive Control: Human brain lysate. Western blot: tissue lysate should be used at 10-50 µg/lane.
Sphingosine 1-phosphate (S1P) is a highly bioactive lipid that has a myriad of biological effects both intracellularly as a second messenger and extracellularly by binding to the S1P(1-5)/Gprotein- coupled receptors of the endothelial differentiation gene family. Intracellularly, at least two enzymes, sphingosine kinase (1 and 2) and S1P phosphatase, regulate the activity of S1P by governing the phosphorylation of S1P. SPPase2 is localized to the endoplasmic reticulum. The enzymatic activity and localization of SPPase2 are similar to SPPase1. The tissue expression of SPPase2 was different from that of SPPase1. SPPase2 is an important member of the SPPase family whichplay a role in attenuating intracellular S1P signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ39004; sphingosine 1-phosphate phosphohydrolase 2; Sphingosine-1-phosphatase 2; Sphingosine-1-phosphate phosphatase 2; sphingosine-1-phosphate phosphotase 2; SPPase2
Gene Aliases: SGPP2; SPP2; SPPase2
UniProt ID: (Human) Q8IWX5
Entrez Gene ID: (Human) 130367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support