Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
This antibody is predicted to react with monkey based on 100% sequence homology.
TMEM97, or MAC30, is a conserved transmembrane protein that helps regulate cellular cholesterol levels through colocalization with NPC1. TMEM97 has also been linked to several cancers including breast, pancreatic, colorectal and ovarian cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAC30; Meningioma-associated protein 30; S2R; Sigma intracellular receptor 2; Sigma-2 receptor; Transmembrane protein 97
Gene Aliases: 1810014L12Rik; AI115531; AL022956; D11Bhm182e; MAC30; S2R; TMEM97
UniProt ID: (Human) Q5BJF2, (Mouse) Q8VD00
Entrez Gene ID: (Human) 27346, (Mouse) 69071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support