Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Contact Us
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • UBXD5 Antibodies

            Proteintech

            UBXN11 Polyclonal Antibody

            1 Reference
            View all (5) UBXD5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite UBXN11 Polyclonal Antibody

            • Antibody Testing Data (10)
            Application
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 10

            UBXN11 Antibody (13109-1-AP) in IHC (P)

            Immunohistochemical analysis of paraffin-embedded human testis tissue slide using (Product # 13109-1-AP) (UBXN11 Antibody) at dilution of 1:200 (under 10x lens).
            View Product

            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            UBXN11 Antibody in Western Blot (WB)
            UBXN11 Antibody in Western Blot (WB)
            UBXN11 Antibody in Immunoprecipitation (IP)

            Product Details

            13109-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            UBXN11 Fusion Protein Ag3772 (1-324 aa encoded by BC038106)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.47 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MSSPLASLS KTRKVPLPSE PMNPGRRGIR IYGDEDEVDM LSDGCGSEEK ISVPSCYGGI GAPVSRQVPA SHDSELMAFM TRKLWDLEQQ VKAQTDEILS KDQKIAALED LVQTLRPHPA EATLQRQEEL ETMCVQLQRQ VREMERFLSD YGLQWVGEPM DQEDSESKTV SEHGERDWMT AKKFWKPGDS LAPPEVDFDR LLASLQDLSE LVVEGDTQVT PVPGGARLRT LEPIPLKLYR NGIMMFDGPF QPFYDPSTQR CLRDILDGFF PSELQRLYPN GVPFKVISCS SNHTFGDFEG SGDTQPPQTP PILRGSRCTP WGTPV (1-324 aa encoded by BC038106)

            Target Information

            This gene encodes a protein with a divergent C-terminal UBX domain. The homologous protein in the rat interacts with members of the Rnd subfamily of Rho GTPases at the cell periphery through its C-terminal region. It also interacts with several heterotrimeric G proteins through their G-alpha subunits and promotes Rho GTPase activation. It is proposed to serve a bidirectional role in the promotion and inhibition of Rho activity through upstream signaling pathways. The 3' coding sequence of this gene contains a polymoprhic region of 24 nt tandem repeats. Several transcripts containing between 1.5 and five repeat units have been reported. Multiple transcript variants encoding different isoforms have been found for this gene.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Colorectal tumor-associated antigen COA-1; colorectal tumor-associated antigen-1; Socius; UBX domain containing 5; UBX domain-containing protein 11; UBX domain-containing protein 5

            View more View less

            Gene Aliases: 4930506L07Rik; COA-1; D4Bwg1540e; PP2243; SOC; SOCI; UBXD5; UBXN11

            View more View less

            UniProt ID: (Human) Q5T124, (Rat) Q8R512, (Mouse) Q9D572

            View more View less

            Entrez Gene ID: (Human) 91544, (Rat) 192207, (Mouse) 67586

            View more View less

            Function(s)

            protein binding ubiquitin binding molecular_function protein-binding activity modulator

            Process(es)

            proteasome-mediated ubiquitin-dependent protein catabolic process biological_process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            • Social Media
            • Contact Us
            • Report a Site Issue
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Argentina flag icon
            Argentina

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-7vhdw:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0