Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: AKKLDKENL SDERASGQSC TLPKRSDSEL KDEKPKRPED ESGRDYRERE REYERDQEHI LRERERLKRQ EEERRRQKER YEKEKTFKRK EEEMKKEKDT LRDKGKKAES TESIGSSEKT EKKEEVVKRD RIRNKDRPAM QLYQPGARSR NRLCPPDDST KSGDSAAERK QESGISHRKE GGEE (301-483 aa encoded by B C121017)
UPF3B is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The protein is one of two functional homologs to yeast Upf3p. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome X. Two splice variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hUPF3B; hUpf3p-X; mental retardation, X-linked 62; Nonsense mRNA reducing factor 3B; Regulator of nonsense transcripts 3B; Up-frameshift suppressor 3 homolog B; Up-frameshift suppressor 3 homolog on chromosome X; UPF3B pseudogene 1; UPF3B pseudogene 2; UPF3B pseudogene 3
Gene Aliases: 5730594O13Rik; AI317193; AW541158; HUPF3B; MRX62; MRXS14; RENT3B; RGD1560264; UPF3B; UPF3BP1; UPF3BP2; UPF3BP3; Upf3p-X; UPF3X
UniProt ID: (Human) Q9BZI7
Entrez Gene ID: (Human) 65109, (Rat) 313449, (Mouse) 68134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support