Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 22; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific protease 22; Ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; Ubiquitin-specific-processing protease 22
Gene Aliases: KIAA1063; USP22; USP3L
UniProt ID: (Human) Q9UPT9
Entrez Gene ID: (Human) 23326
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support