Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 5
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; Cellular nucleic acid-binding protein; CNBP; erythroid differentiation-related; HGNC:13164; sterol regulatory element-binding protein; zinc finger protein 273; Zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein)
Gene Aliases: AA408710; CNBP; CNBP1; DM2; PROMM; RNF163; ZCCHC22; ZNF9
UniProt ID: (Human) P62633, (Mouse) P53996
Entrez Gene ID: (Human) 7555, (Mouse) 12785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support