Whether you are at the discovery phase or ready to commercialize, Thermo Fisher Scientific has integrated, end-to-end solutions that can scale with you no matter what stage you are at in the gene therapy workflow. Use the tool below to explore our end-to-end gene therapy solutions that help you deliver life-changing therapies.
Looking for cell therapy products? Search and filter cell therapy products, services, and solutions
Contact a specialist
More Filters
View all, or filter by any of the categories below to find resources that are relevant to you
Product Name
Description
The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent
as well as a transfection booster and production enhancer
which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.
Gibco CTS Viral Production Medium is a chemically defined
serum-free
protein-free
animal-origin-free
and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System. CTS Viral Production Medium is specifically developed for growth and transfection of CTS Viral Production Cells 2.0 (suspension-adapted HEK293F-derived cells) for adeno-associated virus (AAV) production.
Gibco CTS Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F cell line and are a core component of the CTS AAV-MAX Helper-Free AAV Production System. The cells have been adapted to the chemically defined CTS Viral Production Medium and are maintained in suspension culture in that medium. CTS Viral Production Cells 2.0 can be transfected using the CTS AAV-MAX Transfection Kit for adeno-associated virus (AAV) production.
Gibco CTS Viral-Plex Complexation Buffer is a chemically degined
protein-free
phenol-red-free buffer for complexing plasmic DNA with a transfection reagent for standard transfection applications. It is a part of the CTS AAV-MAX system and is designed to be used with the CTS AAV-MAX Transfection Kit for the transient transfection of CTS Viral Production Cells 2.0.
Gibco Viral Production Medium
AGT
is the dry-format version of CTS Viral Production Medium
a chemically defined
serum-free
protein-free
animal origin-free
phenol red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production.
The Gibco AAV-MAX Helper-Free AAV Production System is a scalable
high-titer platform for suspension AAV production.
The Gibco AAV-MAX Transfection Kit is an essential component of the AAV-MAX Helper-Free AAV Production System. It is formulated for the transient transfection of Viral Production Cells 2.0.
Gibco Viral Production Medium is a chemically defined
serum-free
protein-free medium
that is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells) for adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System.
Gibco Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System.
Gibco Viral-Plex Complexation Buffer is a chemically defined
protein-free
phenol red-free buffer for complexing plasmid DNA with transfection reagent for standard transfection applications.