The Thermo Scientific Pegasus ESD Test System is a low parasitic 2-pin tester used to characterize protection structures at wafer, die, or package level. The Pegasus can also be used as part of your device qualification routine, providing supplemental data to the results gathered when using an automated (relay based) high pin count ESD test system.  Testing is available to the latest industry standards for Human Body Model (HBM), Machine Model (MM) waveforms, as well as offering a true System Level (150pF/330Ω) IEC like waveform based on the EOS/ESD Association’s Human Metal model (HMM). The system’s pulse source design and pulse source delivery method ensures waveform fidelity directly at the device under test, not only at the generator output. Current waveforms can be automatically captured and analyzed for each ESD event.  In addition, Voltage waveforms can be captured and used to determine the turn-on level of protection structures. They can also be used as a means of failure determination, as the voltage waveforms will generally show changes after ESD events.
Key Features
Fully compliant testing
Fully compliant, Human Body Model (HBM) and Machine Model (MM) testing per the latest industry standards.
System level ESD
True system level ESD 150pF/330Ω network (not a 50Ω emulator) available to meet the proposed Human Metal Model (HMM) requirements.
Wafer probes
2 pin connection via wafer probes to any device in a package, die, or wafer.
Failure analysis
Pre- and post-curve trace measurements for comparative failure analysis.
Oscilloscope interface
Oscilloscope interface (GPIB, USB or TCP/IP) with automatic internal attenuation.
Storage
Storage of all waveforms with automatic analysis using the latest industry standards.
ESD waveform generators
Pods (ESD waveform generators) can easily be installed on a prober station.
Supply controls
Bias supply controls for pre- and post-Idd measurements.
Specifications
Style Sheet for Products Table Specifications
Waveforms
  • HMM - Proposed ESDA HMM; HBM - JEDEC/ESDA JS-001, MIL-STD 883, and AEC Q100-002; MM - ESDA STM5.2, JEDEC JESD22-A115, and AEC Q100-003
ZAP Voltage
  • +/- 15 V to 12 kV for HMM and HBM and +/- 50 V to 2 kV for MM 
ZAP Voltage Resolution
  • 1V
ZAP Voltage Accuracy
  • 1% of set point +/-5 V source performance ZAP  
ZAP count/interval range
  • 1 to 9999 count / 0.3 to 10 second intervals
Curve Tracing
  • Voltage: 0 to 200 V (consult factory for higher voltages)
  • Resolution: 10 mV
  • Voltage Ranges: 200 mV, 2 V, 20 V, 200 V
  • Current: 0 to 1 A
  • Resolution: 100 nA
  • Current Ranges: 1 μA, 10 μA, 100 μA, 1 mA, 10 mA, 100 mA, 1 A Accuracy 0.5% of range
  • Output: 22 W: 21 V @ 1 A to 210 V @ 0.1 A
  • Resistance Detection: 1 ohm to 200 M ohm
  • Measurment Points: 10 to 250
Pass/ Fail Criteria 
  • Relative and Absolute Values
Main Controller Dimension
  • 43.2 cm (17 in) W x 43.2 cm (17 in) D x 13.2 cm (5.2 in) H 
Low vacuum mode
  • 16.4 cm (6 in) W x 12.7 cm (5 in) D x 8.9 cm (3.5 in) H
Weight
  • Controller: 8.4 Kg (18.5 lbs)
  • Pod: 1.4 Kg (3 lbs)
Main Controller Power
  • 100 to 240 VAC, 2 Amp, 50/60 Hz
Temperature Range
  • Operating Temperature: +15°C to +40°C (+59°F to +104°F)
  • Non-operating temperature: 4°C to +60°C (+40°F to +140°F)
  • Humidity Range: 30-60% non-condensing
Power Requirements
  • System: 100-240 VAC, 2A, 50/60 Hz
  • Computer and Monitor: 100-240 VAC, 6.5A, 50/60 Hz 
Applications
esd_thumb_274x180_144dpi
ESD Semiconductor Qualification
Every electrostatic discharge (ESD) control plan is required to identify devices that are sensitive to ESD. We offer a complete suite of test systems to help with your device qualification requirements.
Style Sheet for Komodo Tabs
Techniques
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
ESD Compliance Testing
Electrostatic discharge (ESD) can damage small features and structures in semiconductors and integrated circuits. We offer a comprehensive suite of test equipment which verifies that your devices meet targeted ESD compliance standards.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards