cd49 - Key/Resources/Appl/Tech/Doc/Contact
Fiber analysis software
Direct observation and measurement of micro and nano fibers are faster, more efficient and easier than ever before, with improved Thermo Scientific Phenom FiberMetric Software. 
In combination with the Phenom Desktop SEM, the FiberMetric Software allows you to produce accurate size information from micro- and nano-fiber samples.
Fiber characterization
Automated fiber characterization generates hundreds of measurements in seconds. Not only does this result in faster data, but it also means that FiberMetric Software provides more consistent observations when compared to manual measurements.
With FiberMetric Software, it has even become possible to measure and analyze samples with large fiber diameter differences.
Key Features
Faster data collection
Fast and automated collection of all statistical data.
Accurate
View and measure micro and nano fibers with unmatched accuracy.
Save time
Save time with automated measurements.
Specifications
Style Sheet for Products Table Specifications
Fiber Detection
  • 100 nm to 40 μm
  • 1 to 1000 measurements per image
Output
  • XML-data file (incl. diameter measurements and pore surface areas)
  • JPG, TIFF
  • Max. 1024 x 1024 pixel image
  • Customized fiber and pore distribution histogram
  • Minimum, maximum and average fiber size
  • Standard deviation
  • Fiber orientation
Part of ProSuite Software
  • Network storage enabled
  • Phenom integrated system 
Style Sheet for Komodo Tabs
Resources
Video Player is loading.
Current Time 0:00
Duration 1:27
Loaded: 11.39%
Stream Type LIVE
Remaining Time 1:27
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Customer testimonal from Bas de Laat, Synapse Research Institute.
    Video Player is loading.
    Current Time 0:00
    Duration 1:27
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 1:27
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Customer testimonal from Bas de Laat, Synapse Research Institute.
      Style Sheet to change H2 style to p with em-h2-header class
      Contact us
      Electron microscopy services for
      the materials science
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards