STEC testing in high-risk foods
STEC bacteria are primarily found in the intestines of ruminant animals, such as cattle, sheep, and goats. These animals can carry the bacteria without showing any symptoms of illness. STEC can contaminate a wide range of foods, both raw and processed. 
STEC testing applications
STEC testing for dairy
STEC testing in dairy plays a crucial role in ensuring the safety and quality of products. Raw milk and dairy products made from raw milk (such as camembert) are often associated with STEC infection, as they have not undergone the process of pasteurization (which eliminates or reduces harmful bacteria). Implementing stringent testing measures, along with proper sanitation practices and adherence to industry regulations, is therefore essential to mitigate the risk of contamination.
STEC testing for meat
Given that STEC can be present in the intestines of animals, particularly cattle and other grass-eating populations, testing meat products helps identify potential contamination.
Specifically, ground beef is frequently associated with outbreaks as the grinding process can introduce bacteria from the surface of the meat throughout the product, increasing the risk of cross-contamination. This means that even if the exterior of a ground beef product is cooked to a safe temperature, the interior may not reach the same temperature, allowing bacteria to survive.
By implementing robust testing protocols, meat producers and regulatory agencies can ensure safety and take appropriate measures to prevent outbreaks and protect public health.
STEC testing for fresh produce
STEC testing in fresh produce, particularly with a focus on leafy greens, is of the utmost importance in ensuring food safety. Food items, such as lettuce and spinach, have been linked to several outbreaks, as their porous nature and close-to-ground growth environment make them more likely to come into contact with contaminated soil, water, or animal feces. This, coupled with their usually raw consumption, increases the chance of ingesting bacteria. Industry stakeholders must work together to implement preventive measures and increased testing.
STEC environmental monitoring
STEC testing within the production environment in the food industry is crucial for ensuring the safety and quality of food products. Food may be contaminated during various stages of production, processing, and distribution. Testing the production environment involves sampling surfaces, equipment, water sources, and utensils in food production facilities to detect the presence of STEC. This testing helps identify potential sources of contamination, assesses the effectiveness of sanitation practices, and allows for implementation of necessary corrective actions, giving consumers the confidence that their food is prepared safely.
STEC testing for poultry
While poultry is not as commonly associated with STEC contamination as ground beef, it can still be a potential source of infection if not handled or cooked properly. Poultry can become contaminated with STEC during various stages, including production, processing, and handling. This can occur through contact with contaminated surfaces, equipment, or cross-contamination from other sources. Proper hygiene practices, such as avoiding cross-contamination, following good sanitation procedures, and implementing stringent quality control measures throughout the poultry production chain, are crucial in reducing risk.  
For Research Use Only. Not for use in diagnostic procedures.