EVOS Vessel Holders and Stage Plates
EVOS vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides. Custom configurations are also available upon request.
 Download a printable guide to EVOS vessel holders and stage plates
AMEPVH001
 AMEPVH002 AMEPVH003
AMEPVH001
Holds two 25 mm x 75 mm standard microscope slides, chamber slides, etc
AMEPVH002
Holds four 35 mm petri dishes
AMEPVH003
Holds two 60 mm petri dishes
AMEPVH004 AMEPVH005 AMEPVH006
AMEPVH004
Holds one 100 mm petri dish
AMEPVH005
Holds two 25 cm2 flasks; rectangular or triangular
AMEPVH006
Holds one Nunc® T-75 flask; 75 cm2
 AMEPVH007    AMEPVH010
AMEPVH007
Holds one hemocytometer
AMEPVH009
Universal stage insert
AMEPVH010
Holds one BD/Greiner T-25 flask; 25 cm2
 AMEPVH010  AMEPVH012  AMEPVH013
Holds one Nunc®/SPL IVF 4-well dish 
AMEPVH012
Holds one SPL T-75 flask; 75 cm2
AMEPVH013
Holds four Ibidi® 35 mm petri dishes
 AMEPVH014  AMEPVH017  AMEPVH018
Holds two Ibidi® 50 mm petri dishes 
AMEPVH017
Holds one KOVA® Glasstic® slide 10
AMEPVH018
Holds one Nunc® T-25 flask; 25 cm2
AMEP4686 AMEP4691 AMEP4692 
Stage plate for multiwell vessels; also holds one Corning® T-75 flask 
Stage plate with 110 mm x 160 mm opening (use with AMEP4692 for standard sizes)
Stage plate adaptor with 110 mm x 160 mm opening for standard size
AMEPVH021
 AMEPVH022  AMEPVH022
Securely holds two 25 mm x 75 mm standard microscope slides, chamber slides, etc.
AMEPVH022
Intermediate plate for automated stage; securely holds multi-well vessels with convenient lever adaptor for AMEPVH001 and AMEPVH009
AMEPVH023
Holds multi-well vessels. Adaptor for AMEPVH001 and AMEPVH009
AMEPVH001
 AMEPVH002 AMEPVH003
AMEPVH028
Well plate for incubator
AMEPVH029
Holds one 35mm Petri dish
AMEPVH030
Holds two 35mm Petri Dishes
AMEPVH004  AMEPVH006
AMEPVH031
Holds one 60mm Petri dish
AMEPVH032
Holds two 60mm Petri dishes
 AMEPVH033
Holds one T-25 flask
AMEPVH007  
AMEPVH034
Holds two T-25 flasks
AMEPVH001
 AMEPVH002 AMEPVH003
AMEPVH001
Holds two 25 mm x 75 mm standard microscope slides, chamber slides, etc
AMEPVH002
Holds four 35 mm petri dishes
AMEPVH003
Holds two 60 mm petri dishes
AMEPVH004 AMEPVH005 AMEPVH006
AMEPVH004
Holds one 100 mm petri dish
AMEPVH005
Holds two 25 cm2 flasks; rectangular or triangular
AMEPVH006
Holds one Nunc® T-75 flask; 75 cm2
 AMEPVH007    AMEPVH010
AMEPVH007
Holds one hemocytometer
AMEPVH009
Universal stage insert
AMEPVH010
Holds one BD/Greiner T-25 flask; 25 cm2
 AMEPVH010  AMEPVH012  AMEPVH013
Holds one Nunc®/SPL IVF 4-well dish 
AMEPVH012
Holds one SPL T-75 flask; 75 cm2
AMEPVH013
Holds four Ibidi® 35 mm petri dishes
 AMEPVH014  AMEPVH017  AMEPVH018
Holds two Ibidi® 50 mm petri dishes 
AMEPVH017
Holds one KOVA® Glasstic® slide 10
AMEPVH018
Holds one Nunc® T-25 flask; 25 cm2
AMEP4686 AMEP4691 AMEP4692 
Stage plate for multiwell vessels; also holds one Corning® T-75 flask 
Stage plate with 110 mm x 160 mm opening (use with AMEP4692 for standard sizes)
Stage plate adaptor with 110 mm x 160 mm opening for standard size
AMEPVH021
 AMEPVH022  AMEPVH022
Securely holds two 25 mm x 75 mm standard microscope slides, chamber slides, etc.
AMEPVH022
Intermediate plate for automated stage; securely holds multi-well vessels with convenient lever adaptor for AMEPVH001 and AMEPVH009
AMEPVH023
Holds multi-well vessels. Adaptor for AMEPVH001 and AMEPVH009
AMEPVH001
 AMEPVH002 AMEPVH003
AMEPVH028
Well plate for incubator
AMEPVH029
Holds one 35mm Petri dish
AMEPVH030
Holds two 35mm Petri Dishes
AMEPVH004  AMEPVH006
AMEPVH031
Holds one 60mm Petri dish
AMEPVH032
Holds two 60mm Petri dishes
 AMEPVH033
Holds one T-25 flask
AMEPVH007  
AMEPVH034
Holds two T-25 flasks
Ordering Information
Catalog # Name Size Price (USD) Qty
AMEP4684 EVOS™ Stage Plate for Tokai Hit Heating Tray 1 each -
AMEP4685 EVOS™ Stage Plate for BioFlux Heating Stage 1 each -
AMEP4686 EVOS™ Stage Plate for Multi-well Vessels or T-75 Flask 1 each -
AMEP4687 EVOS™ Stage Plate for Multi-well Vessels, max objective clearance Each
440.00
AMEP4691 EVOS™ Stage Plate, 160 x 110 mm opening 1 each -
AMEP4692 EVOS™ Stage Plate Adapter Each
455.00
AMEP4619 EVOS™ Vessel Holders Pack Each
1,268.00
AMEPVH001 EVOS™ Vessel Holder, two 25 x 75 mm slides Each
145.00
AMEPVH002 EVOS™ Vessel Holder, four 35-mm Petri dishes Each
145.00
AMEPVH003 EVOS™ Vessel Holder, two 60-mm Petri dishes Each
146.00
AMEPVH004 EVOS™ Vessel Holder, one 100-mm Petri dish Each
145.00
AMEPVH005 EVOS™ Vessel Holder, two 25-cm T-flasks Each
145.00
AMEPVH006 EVOS™ Vessel Holder, Nunc™ T-75 flask Each
147.00
AMEPVH007 EVOS™ Vessel Holder, Hemocytometer Each
145.00
AMEPVH008 EVOS™ Vessel Holder, Greiner T-75 flask Each
146.00
AMEPVH009 EVOS™ Vessel Holder, universal Each
180.00
AMEPVH010 EVOS™Vessel Holder, Greiner/BD T-25 flask Each
146.00
AMEPVH011 EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish Each
144.00
AMEPVH012 EVOS™ Vessel Holder, SPL T-75 flask Each
146.00
AMEPVH013 EVOS™ Vessel Holder, ibidi 35 mm Petri dishes Each
146.00
AMEPVH014 EVOS™ Vessel Holder, ibidi 50 mm Petri dishes Each
145.00
AMEPVH017 EVOS™ Vessel Holder, KOVA™ Glasstic™ slide 10 Each
435.00
AMEPVH018 EVOS™ Vessel Holder, one Nunc™ T-25 flask Each
359.00
AMEPVH021 EVOS™ Vessel Holder for Automated Stage, two 25 x 75 mm slides Each
467.00
AMEPVH022 EVOS™ Vessel Holder/Adapter for Automated Stage, with retention, multi-well plates Each
461.00
AMEPVP01 EVOS™ Vessel Plate for Large Vessels Each
152.00
AMEPVH028 EVOS™ Onstage Vessel Holder, Multi-Well Plates Each
440.00
AMEPVH029 EVOS™ Onstage Vessel Holder, One 35 mm Petri Dish Each
430.00
AMEPVH030 EVOS™ Onstage Vessel Holder, Two 35 mm Petri Dish Each
430.00
AMEPVH031 EVOS™ Onstage Vessel Holder, one 60-mm Petri dish Each
440.00
AMEPVH032 EVOS™ Onstage Vessel Holder, two 60-mm Petri dishes Each
430.00
AMEPVH033 EVOS™ Onstage Vessel Holder, One 25 mL Flask Each
440.00
AMEPVH034 EVOS™ Onstage Vessel Holder, Two 25 mL Flask Each
430.00
AMEPVH037 EVOS™ Onstage Vessel Holder, one 100-mm Petri dish Each
425.00
For Research Use Only. Not for use in diagnostic procedures.