Flow Cytometry Instruments
Flow cytometry cell analyzer and cell sorter
Explore our portfolio of Attune Flow Cytometers for high-speed, high-clog resistance, automation-adopted cell analysis and the Bigfoot Spectral Cell Sorter for precision high-speed cell sorting. All our flow cytometry instruments offer flexibility in optical configuration, true high-throughput capabilities, and exceptional speed. 
Attune Xenith Flow Cytometer
Fast, flexible, and spectrally brilliant—simplify high-speed, high-parameter analysis.
Attune Cytpix Flow Cytometer
Imaging-enhanced flow cytometer that includes a high-speed brightfield camera for simultaneous morphological analysis.
Attune NxT Flow Cytometer
Hardware features enable exceptionally fast sample flow rates, high clog resistance, and outstanding reliability.
Bigfoot Spectral Cell Sorter
Up to 9 lasers and 60 detectors, remarkably fast multi-way plate sorting, and easy-to-use flexible design.
Solutions for scale up cell analysis
Autosamplers, robotic automation options, and data integration software can streamline your workflow and increase throughput. Other experimental needs will include liquids, bottles, beads, and other consumables. 
Autosamplers
The walkaway automation system seamlessly integrates with your Attune Flow Cytometer for increased productivity.
Robotic automation
Extend your unmanned runtime settings and scalability with robotic integration applications for the Attune Flow Cytometers.
Data integration software
Dynamic scheduling in the Momentum Workflow Scheduling Software allows users to successfully create multiple workflows.
These resources can help you increase your understanding of the fundamentals and scientific principles of flow cytometry and expand your abilities for specialty applications and techniques.
Educational resources
Flow Cytometry Panel Design Service & Panel Builder Tool
Immunology at Work Resource Center
Antibodies for Flow Cytometry
Antibodies for Flow Cytometry
Register for flow webinar series
Cy™  is a trademark or registered trademark of GE Healthcare. 
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. 
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and is used under license. 