Normalize and troubleshoot your qPCR reactions
ROX is a fluorescent passive reference dye with a level of fluorescence that does not change during the PCR process. Normalization of qPCR data using ROX reduces the influence of well-to-well variability such as uneven illumination, slight variation in optics, and differences in the amount of condensation.
Most real-time PCR instruments use ROX to normalize for these and other sources of variability. ROX is the passive reference dye of choice because it does not influence the PCR process and its emission spectrum does not influence those of many other TaqMan reporter and quencher dyes.
Features
Features of ROX Passive Reference Dye include:
Normalizes non-PCR-related fluctuations in fluorescence
Using a passive reference like ROX dye to normalize data helps achieve a higher level of precision among technical replicates. Without normalization, more replicates may be required to achieve comparable precision levels, increasing the time and resources required for each run. Data normalized with ROX dye is simply more precise than data without it, enabling researchers to run fewer replicates while maintaining statistical power.
Provides a stable baseline for multiplex quantitative PCR and RT-PCR
In a multicomponent plot, the ROX signal should remain constant through the qPCR process. Any non-PCR related events will lead to spikes, dips, or other abnormal behavior in the reporter and ROX signals.
Using ROX dye to troubleshoot common qPCR issues
Unlike probe-linked dyes that allow for detection of one or more targets in the same well, passive reference dyes are not influenced by the PCR process and can be used to normalize against several variables that create well-to-well variation.
Common sources of process errors that can compromise your data
  • Bubbles in wells
  • Evaporation
  • Condensation or droplets
  • Instrument issues, such as electrical surges
Illustration of common sources of errors that can compromise qPCR data
Monitoring ROX fluorescence in a Multicomponent Plot enables researchers to detect various quality control issues in a qPCR run. To  troubleshoot your data using the Multicomponent Plot, review the shape of the ROX dye signal:
If the ROX dye reading …
Possible issues
Remains flat throughout run
Expected result. ROX fluorescence should remain constant for the duration of the reaction because it is not affected by amplification as shown in the figure below.
Increases during the run
Evaporation may be occurring because the ROX concentration is increasing as the volume decreases.
Momentarily spikes or drops
Sudden changes are interfering with data recording in that well, such as air bubbles or an electrical surge.
Sample multicomponent plot of expected result using ROX dye
Illustration of common sources of errors that can compromise qPCR data
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Instrument and master mix compatibility
      ROX dye has been qualified for use on all Applied Biosystems real-time PCR Instruments. ROX dye is detected in Dye Channel 4 (excitation filter: 580 ± 10 nm, emission filter: 623 ± 14 nm).
      Dye compatibility with QuantStudio systems
      Channel
      1
      2
      3
      4
      5
      6
      Excitation filter
      470 ± 15 nm
      520 ± 10 nm
      550 ± 10 nm
      580 ± 10 nm
      640 ± 10 nm
      662 ± 10 nm
      Emission filter
      520 ± 15 nm
      558 ± 12 nm
      586 ± 10 nm
      623 ± 14 nm
      682 ± 14 nm
      711 ± 12 nm
      Examples of compatible dyes
      FAM, SYBR Green I, SYTO 9, fluorescein, PTS dye
      VIC, JOT, TET, HEX, Yakima Yellow, SUN
      TAMRA, NED, BODIPY TMR-X, ABY, Cy3
      ROX, Texas Red, JUN
      LIZ, Cy5, Mustang Purple
      Cy5.5, Alexa Fluor 680, TYE 685
      QuantStudio 1 system
      QuantStudio 3 system
      QuantStudio 6 Pro system
      QuantStudio 5, 7 Pro, and 12K Flex systems
      Our standalone ROX kit is compatible with all other TaqMan and TaqPath master mixes and all TaqMan Assays that do not contain JUN, Texas Red, or Alexa 594 as the reporter dye.
      ROX can also be used with competitor master mixes if the mix does not contain a coloring agent or passive reference dye. Optimization will also be necessary.
      Sample data
      amplification-plot11
      amplification-plot22
      How ROX dye normalizes qPCR amplification plots. 
      (A) Without ROX dye normalization, bubbles in wells affect fluorescence readings. (B) When data was normalized with ROX dye as a passive reference, aberrations caused by the bubbles are removed.
      Ordering information
      ROX dye is available as a standalone kit or in a selection of Applied Biosciences qPCR master mixes.
      Catalog # Name Size Price (USD) Qty
      12223012 Each
      69.65
      Online Exclusive
      74.25
      Save 4.60 (6%)
      For Research Use Only. Not for use in diagnostic procedures.
      *TaqPath master mixes are general purpose reagents labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.