Front angle on the ProFlex PCR System, including the user interface home screen
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems thermal cyclers with the flexible configuration and control features that fit how you work today. Interchangeable block formats allow you to maximize your throughput or run independent experiments concurrently. And now you can access your ProFlex PCR System with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods and monitor runs from any smartphone or desktop computer.
Video Player is loading.
Current Time 0:00
Duration 1:16
Loaded: 0%
Stream Type LIVE
Remaining Time 1:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video: ProFlex thermal cyclers to maximize your throughput with flexibility
    Maximize your throughput using Proflex PCR system that supports up to 5 interchangeable blocks based on your needs.
    PCR system with interchangeable blocks for high throughput PCR or simultaneous experiments
    The ProFlex PCR System has five different blocks that can be changed with the flip of a switch, including the first-of-its-kind 3 x 32-well block. This block allows up to three experiments to be run simultaneously, completely independently of each other and without interference. Dual 96-well and dual 384-well blocks are available for your high-throughput PCR sequencing needs, and a dual flat block is available to support Applied Biosystems OpenArray plate technology for genotyping analysis on the Applied Biosystems QuantStudio 12K Flex Real-Time PCR System.
    Top of the ProFlex PCR System containing the 3 x 32-well block
    ProFlex PCR System with the 3 x 32-well block for running up to three independent experiments simultaneously.
    Top and partial screen of the ProFlex PCR System containing the Dual 384-well block
    Dual 384-well block for higher-throughput sequencing using ProFlex PCR Systems.
    Top of the ProFlex PCR System containing the Dual flat block
    Dual flat block on the ProFlex PCR System supports OpenArray plate technology for genotyping analysis on the QuantStudio 12K Flex Real-Time PCR System.
    Top and screen of the ProFlex PCR System containing the 96-well block
    96-well block for standard PCR applications using the ProFlex thermal cycler.
    Top of the ProFlex PCR System containing the Dual 96-well block
    Dual 96-well block for the ProFlex thermal cycler for high-throughput sequencing.
    Stay in control with Thermal Cycler Fleet Control Software
    Diagram of multiple thermal cyclers connected to one tablet to represent Fleet Control Software
    Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
    Learn more about Thermal Cycler Fleet Control Software
    Ordering information
    Catalog # Name Size Price (USD) Qty
    A41182 ProFlex™ PCR System + Plastics Package, 3 x 32-well, 8-tube strips, 1 package Each
    8,322.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,870.00
    Save 5,548.00 (40%)
    A41183 ProFlex™ PCR System + Plastics Package, 3 x 32-well, TriFlex plates, 1 package Each
    8,334.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,890.00
    Save 5,556.00 (40%)
    A41184 ProFlex ™ PCR System + Plastics Package, 1 x 96-well, EnduraPlates, 1 package Each
    8,202.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,670.00
    Save 5,468.00 (40%)
    A41185 ProFlex™ PCR System + Plastics Package, 1 x 96-well, 8-tube strips, 1 package Each
    8,202.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,670.00
    Save 5,468.00 (40%)
    A41186 ProFlex™ PCR System + Plastics Package, 2x96-well, EnduraPlates Each
    13,188.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    21,980.00
    Save 8,792.00 (40%)
    A41187 ProFlex™ PCR System + Plastics Package, 2x384-well, EnduraPlates Each
    12,348.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    20,580.00
    Save 8,232.00 (40%)
    Catalog # Name Size Price (USD) Qty
    A36396 ProFlex™ PCR System Extended Warranty + Plastics Package, REX, 3x32-well, 1 package Each
    8,012.32
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,353.86
    Save 5,341.54 (40%)
    A36958 ProFlex™ PCR System Extended Warranty + Plastics Package, ABRC, 3x32-well, 1 package Each
    8,153.68
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,589.46
    Save 5,435.78 (40%)
    A36959 ProFlex™ PCR System Extended Warranty + Plastics Package, REX, 1 x 96-well, 1 package Each
    7,697.71
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,829.51
    Save 5,131.80 (40%)
    A36960 ProFlex™ PCR System Extended Warranty + Plastics Package, ABRC, 1 x 96-well, 1 package Each
    7,832.23
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,053.71
    Save 5,221.48 (40%)
    A36961 ProFlex™ PCR System Extended Warranty + Plastics Package, REX, 2 x 96-well, 1 package Each
    11,933.95
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    19,889.91
    Save 7,955.96 (40%)
    A36962 ProFlex™ PCR System Extended Warranty + Plastics Package, ABRC, 2 x 96-well, 1 package Each
    12,775.27
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    21,292.11
    Save 8,516.84 (40%)
    A36963 ProFlex™ PCR System Extended Warranty + Plastics Package, REX, 2 x 384-well, 1 package Each
    11,156.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    18,594.16
    Save 7,437.66 (40%)
    A36964 ProFlex™ PCR System Extended Warranty + Plastics Package, ABRC, 2 x 384-well, 1 package Each
    11,377.66
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    18,962.76
    Save 7,585.10 (40%)
    Catalog # Name Size Price (USD) Qty
    4484075 ProFlex™ 96-well PCR System, 1 instrument Each
    8,762.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,480.00
    Save 4,718.00 (35%)
    4484073 ProFlex™ 3 x 32-well PCR System, 1 instrument Each
    8,885.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    13,670.00
    Save 4,784.50 (35%)
    4484076 ProFlex™ 2 x 96-well PCR System, 1 instrument Each
    13,858.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    21,320.00
    Save 7,462.00 (35%)
    4484078 ProFlex™ 2 x Flat PCR System, 1 instrument Each
    13,357.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    20,550.00
    Save 7,192.50 (35%)
    4484077 ProFlex™ PCR System, 2 x 384-well, 1 instrument Each
    13,422.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    20,650.00
    Save 7,227.50 (35%)
    Catalog # Name Size Price (USD) Qty
    4483637 ProFlex™ 96-well Sample Block, 1 unit Each
    3,142.75
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    4,835.00
    Save 1,692.25 (35%)
    4483638 ProFlex™ 3 x 32-well Sample Block, 1 unit Each
    4,329.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    6,660.00
    Save 2,331.00 (35%)
    4484071 ProFlex™ 2 x 96-well Sample Block, 1 unit Each
    8,300.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,770.00
    Save 4,469.50 (35%)
    4484074 ProFlex™ 2 x flat Sample Block, 1 unit Each
    8,144.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,530.00
    Save 4,385.50 (35%)
    4484072 ProFlex™ 2 x 384-well Sample Block, 1 block Each
    7,995.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,300.00
    Save 4,305.00 (35%)
    High-performance PCR and qPCR plastics for optimal results
    Five multi-colored PCR tube plates
    Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
    Explore online PCR and qPCR plastics selection tool
    Resources
    Thermal Cycler Software and Firmware Files
    Download the latest updates for your instrument
    Interested in private label thermal cyclers?
    Submit an inquiry
    Support
    Service
    We provide a full range of service options for all Applied Biosystems thermal cyclers.
    View service plans
    Multiplex Dynamic Temperature Verification Service
    Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
    View temperature service options
     Download MDTV flyer
    For Research Use Only. Not for use in diagnostic procedures.