Invitrogen PowerEase Touch Power Supply for protein and nucleic acid gel electrophoresis
From gel electrophoresis and western transfer to more demanding applications, we offer a range of powerful, easy-to-use electrophoresis power supplies. PowerEase Touch Power Supplies can bring a new level of convenience to your electrophoresis experiments. With a bright, LCD touch screen interface, enter in custom methods or use the pre-programmed protocols for Invitrogen protein gels and gel transfers. Get started with the available welcome pack bundles.
Watch video
Electrophoresis chambers Protein gels
Find the appropriate electrophoresis power supply
 Invitrogen PowerEase Touch 120 W Power Supply for protein and nucleic acid gel electrophoresisInvitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresisInvitrogen PowerEase Touch 600WPowerEase Touch HV Power Supply photo
 PowerEase Touch 120W Power SupplyPowerEase Touch 350W Power Supply
PowerEase Touch 600W Power SupplyPowerEase Touch HV Power Supply
ApplicationsRunning 1–4 mini gels or 1–2 midi gels, or transferring 1–2 mini gels, RNA/DNA electrophoresisRunning up to 12 mini or 8 midi, or transferring up to 8 mini or 4 midi gels, RNA/DNA electrophoresisRunning up to 16 mini or 14 midi, or transferring up to 8 mini or 4 midi gels, IEF gels, RNA/DNA electrophoresisRunning 1–4 mini gels or 1–2 midi gels, or transferring 1–2 mini gels, ZOOM IPG Strips, IEF gels, RNA/DNA electrophoresis
Number of outputs4444
Output voltage range2–300 V2–300 V2–500 V20 V–3500 V
Output current range1–500 mA1 mA–3A1 mA–3A0.05 mA–500 mA
Power120 W350 W600 W250 W
Modes
  • Constant voltage
  • Constant current
  • Constant power
  • Constant voltage
  • Constant current
  • Constant power
  • Constant voltage
  • Constant current
  • Constant power
  • Constant voltage
  • Constant current
  • Constant power
  • Voltage Ramp
TimerUp to 999 min per stepUp to 999 min per stepUp to 999 min per stepUp to 999 min per step
DisplayBacklit touch screenBacklit touch screenBacklit touch screenBacklit touch screen
Programmable
  • Store up to 100 run methods
  • Program up to 20 steps per method
  • Store up to 100 run methods
  • Program up to 20 steps per method
  • Store up to 100 run methods
  • Program up to 20 steps per method
  • Store up to 100 run methods​
  • Program up to 20 steps per method
Features
  • User adjustable settings for beep volume, screen brightness, sleep mode duration
  • Pop-up feet to adjust viewing angle
  • Safety features include:
    • No load
    • Over temperature
    • Over voltage
    • Over current
    • Load change
    • Ground leak detection
  • User adjustable settings for beep volume, screen brightness, sleep mode duration
  • Pop-up feet to adjust viewing angle
  • Safety features include:
    • No load
    • Over temperature
    • Over voltage
    • Over current
    • Load change
    • Ground leak detection
  • User adjustable settings for beep volume, screen brightness, sleep mode duration
  • Pop-up feet to adjust viewing angle
  • Safety features include:
    • No load
    • Over temperature
    • Over voltage
    • Over current
    • Load change
    • Ground leak detection
  • User adjustable settings for beep volume, screen brightness, sleep mode duration
  • Pop-up feet to adjust viewing angle
  • Safety features include:
    • No load
    • Over temperature
    • Over voltage
    • Over current
    • Load change
    • Ground leak detection
    • Ability to turn off No-Load detection for low current applications
Invitrogen PowerEase Touch 120 W Power Supply for protein and nucleic acid gel electrophoresis
Invitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresis
Invitrogen PowerEase Touch 600W
PowerEase Touch HV
PowerEase Touch Power Supplies feature a modern stackable design and a large touch screen that is easy to operate and program while delivering precise outputs for voltage, current, and power.
Ideal for applications like DNA/RNA electrophoresis, SDS-PAGE, and native PAGE, the 120W, 350W, and 600W models come pre-programmed with protocols for electrophoresis for Invitrogen precast protein gels and are easily programmed for other applications. The 350W model provides enough power to transfer midi size protein gels and the 600W can run IEF gels. The HV model preprogrammed protocols support Invitrogen IEF gels and ZOOM IPG Strips, and is suitable for 2D gel electrophoresis with the ZOOM IPGRunner System.
Features of PowerEase Touch Power Supplies
  • Touch screen LCD display—easily program with clear menu prompts and view run progress
  • Pre-programmed modes for Invitrogen gels & applications—help save time and minimize errors
  • Easy programming and storage of multi-step methods—streamline your processes
  • Precise outputs for voltage, current and power—get reproducible results time after time
  • Modern stackable design—save precious space
  • Pop-up feet—allows users to change screen viewing angle
  • Adjustable device settings—set beep volume, screen brightness, and sleep mode duration to your preference
  • Automatic safety features—no load, over temperature, over voltage, over current, load change and ground leak detection help ensure safety
Video Player is loading.
Current Time 0:00
Duration 0:58
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    PowerEase Touch welcome packs
    PowerEase Touch 120W and 350W Power Supplies are also available in money-saving bundles with a Mini Gel Tank or XCell SureLock Mini-Cell Tank, with or without a blot module. PowerEase Touch HV is also available in combination with the ZOOM IPGRunner System. Use the table below to find the welcome pack that fits your needs.
    PowerEase Touch welcome pack configurations
     Invitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresisMini Gel Tank and Mini Blot ModuleXCell SureLock Mini Cell and Blot ModuleSureLock Tandem Midi Gel Tank and Blot Modules
     PowerEase Touch Power SupplyMini gel tanks and blot modulesMidi gel tank and blot module
    Cat. No.PowerEase Touch
    120 W
    PowerEase Touch
    350 W
    Mini Gel TankMini Blot ModuleXCell SureLock Mini CellXCell SureLock Blot ModuleSureLock Tandem Midi Gel TankSureLock Midi Blot Module
    PSC120M        
    PSC350M        
    PSC120S        
    PSC350S        
    PSC120MB        
    PSC350MB        
    PSC120SB        
    PSC350SB        
    PSC350TT        
    PSC350STMPV*        
    PSC350STMNC**        
    *Also includes SureLock Tandem Midi Pre-cut 0.45 μm PVDF Membranes and Filter Paper, and SureLock Tandem Transfer Tray
    **Also includes SureLock Tandem Midi Pre-cut 0.2 μm Nitrocellulose Membranes and Filter Paper, and SureLock Tandem Transfer Tray
    The PowerEase Touch HV Power Supply and ZOOM IPGRunner Welcome Pack contains all necessary components for first dimension separation. Just add your preferred ZOOM IPG strips and carrier ampholytes.
    Cat. No.PowerEase Touch
    Power Supply
    Other components
    PSHV00ZPowerEase Touch HVZOOM IPGRunner Mini-Cell
    ZOOM IPGRunner Cassettes
    ZOOM Urea
    ZOOM CHAPS
    ZOOM Equilibration Trays
    Power supplies
    Catalog # Name Size Price (USD) Qty
    PS0600 PowerEase™ Touch 600W Power Supply, 115 VAC, 1 each Each
    3,666.65
    Online Exclusive
    3,835.00
    Save 168.35 (4%)
    PSHV00A PowerEase™ Touch HV Power Supply Adapters Each
    110.00
    PSHV00 PowerEase™ Touch HV Power Supply, 115 VAC Each
    3,929.65
    Online Exclusive
    4,115.00
    Save 185.35 (5%)
    PS0120 PowerEase™ Touch 120W Power Supply, 115 VAC, 1 each Each
    951.65
    Online Exclusive
    994.00
    Save 42.35 (4%)
    PS0350 PowerEase™ Touch 350W Power Supply, 115 VAC, 1 each Each
    1,620.65
    Online Exclusive
    1,708.00
    Save 87.35 (5%)
    PowerEase Touch welcome packs
    Find the appropriate electrophoresis power supply
     Invitrogen PowerEase Touch 120 W Power Supply for protein and nucleic acid gel electrophoresisInvitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresisInvitrogen PowerEase Touch 600WPowerEase Touch HV Power Supply photo
     PowerEase Touch 120W Power SupplyPowerEase Touch 350W Power Supply
    PowerEase Touch 600W Power SupplyPowerEase Touch HV Power Supply
    ApplicationsRunning 1–4 mini gels or 1–2 midi gels, or transferring 1–2 mini gels, RNA/DNA electrophoresisRunning up to 12 mini or 8 midi, or transferring up to 8 mini or 4 midi gels, RNA/DNA electrophoresisRunning up to 16 mini or 14 midi, or transferring up to 8 mini or 4 midi gels, IEF gels, RNA/DNA electrophoresisRunning 1–4 mini gels or 1–2 midi gels, or transferring 1–2 mini gels, ZOOM IPG Strips, IEF gels, RNA/DNA electrophoresis
    Number of outputs4444
    Output voltage range2–300 V2–300 V2–500 V20 V–3500 V
    Output current range1–500 mA1 mA–3A1 mA–3A0.05 mA–500 mA
    Power120 W350 W600 W250 W
    Modes
    • Constant voltage
    • Constant current
    • Constant power
    • Constant voltage
    • Constant current
    • Constant power
    • Constant voltage
    • Constant current
    • Constant power
    • Constant voltage
    • Constant current
    • Constant power
    • Voltage Ramp
    TimerUp to 999 min per stepUp to 999 min per stepUp to 999 min per stepUp to 999 min per step
    DisplayBacklit touch screenBacklit touch screenBacklit touch screenBacklit touch screen
    Programmable
    • Store up to 100 run methods
    • Program up to 20 steps per method
    • Store up to 100 run methods
    • Program up to 20 steps per method
    • Store up to 100 run methods
    • Program up to 20 steps per method
    • Store up to 100 run methods​
    • Program up to 20 steps per method
    Features
    • User adjustable settings for beep volume, screen brightness, sleep mode duration
    • Pop-up feet to adjust viewing angle
    • Safety features include:
      • No load
      • Over temperature
      • Over voltage
      • Over current
      • Load change
      • Ground leak detection
    • User adjustable settings for beep volume, screen brightness, sleep mode duration
    • Pop-up feet to adjust viewing angle
    • Safety features include:
      • No load
      • Over temperature
      • Over voltage
      • Over current
      • Load change
      • Ground leak detection
    • User adjustable settings for beep volume, screen brightness, sleep mode duration
    • Pop-up feet to adjust viewing angle
    • Safety features include:
      • No load
      • Over temperature
      • Over voltage
      • Over current
      • Load change
      • Ground leak detection
    • User adjustable settings for beep volume, screen brightness, sleep mode duration
    • Pop-up feet to adjust viewing angle
    • Safety features include:
      • No load
      • Over temperature
      • Over voltage
      • Over current
      • Load change
      • Ground leak detection
      • Ability to turn off No-Load detection for low current applications
    Invitrogen PowerEase Touch 120 W Power Supply for protein and nucleic acid gel electrophoresis
    Invitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresis
    Invitrogen PowerEase Touch 600W
    PowerEase Touch HV
    PowerEase Touch Power Supplies feature a modern stackable design and a large touch screen that is easy to operate and program while delivering precise outputs for voltage, current, and power.
    Ideal for applications like DNA/RNA electrophoresis, SDS-PAGE, and native PAGE, the 120W, 350W, and 600W models come pre-programmed with protocols for electrophoresis for Invitrogen precast protein gels and are easily programmed for other applications. The 350W model provides enough power to transfer midi size protein gels and the 600W can run IEF gels. The HV model preprogrammed protocols support Invitrogen IEF gels and ZOOM IPG Strips, and is suitable for 2D gel electrophoresis with the ZOOM IPGRunner System.
    Features of PowerEase Touch Power Supplies
    • Touch screen LCD display—easily program with clear menu prompts and view run progress
    • Pre-programmed modes for Invitrogen gels & applications—help save time and minimize errors
    • Easy programming and storage of multi-step methods—streamline your processes
    • Precise outputs for voltage, current and power—get reproducible results time after time
    • Modern stackable design—save precious space
    • Pop-up feet—allows users to change screen viewing angle
    • Adjustable device settings—set beep volume, screen brightness, and sleep mode duration to your preference
    • Automatic safety features—no load, over temperature, over voltage, over current, load change and ground leak detection help ensure safety
    Video Player is loading.
    Current Time 0:00
    Duration 0:58
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 0:58
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      PowerEase Touch welcome packs
      PowerEase Touch 120W and 350W Power Supplies are also available in money-saving bundles with a Mini Gel Tank or XCell SureLock Mini-Cell Tank, with or without a blot module. PowerEase Touch HV is also available in combination with the ZOOM IPGRunner System. Use the table below to find the welcome pack that fits your needs.
      PowerEase Touch welcome pack configurations
       Invitrogen PowerEase Touch 350 W Power Supply for protein and nucleic acid gel electrophoresisMini Gel Tank and Mini Blot ModuleXCell SureLock Mini Cell and Blot ModuleSureLock Tandem Midi Gel Tank and Blot Modules
       PowerEase Touch Power SupplyMini gel tanks and blot modulesMidi gel tank and blot module
      Cat. No.PowerEase Touch
      120 W
      PowerEase Touch
      350 W
      Mini Gel TankMini Blot ModuleXCell SureLock Mini CellXCell SureLock Blot ModuleSureLock Tandem Midi Gel TankSureLock Midi Blot Module
      PSC120M        
      PSC350M        
      PSC120S        
      PSC350S        
      PSC120MB        
      PSC350MB        
      PSC120SB        
      PSC350SB        
      PSC350TT        
      PSC350STMPV*        
      PSC350STMNC**        
      *Also includes SureLock Tandem Midi Pre-cut 0.45 μm PVDF Membranes and Filter Paper, and SureLock Tandem Transfer Tray
      **Also includes SureLock Tandem Midi Pre-cut 0.2 μm Nitrocellulose Membranes and Filter Paper, and SureLock Tandem Transfer Tray
      The PowerEase Touch HV Power Supply and ZOOM IPGRunner Welcome Pack contains all necessary components for first dimension separation. Just add your preferred ZOOM IPG strips and carrier ampholytes.
      Cat. No.PowerEase Touch
      Power Supply
      Other components
      PSHV00ZPowerEase Touch HVZOOM IPGRunner Mini-Cell
      ZOOM IPGRunner Cassettes
      ZOOM Urea
      ZOOM CHAPS
      ZOOM Equilibration Trays
      Power supplies
      Catalog # Name Size Price (USD) Qty
      PS0600 PowerEase™ Touch 600W Power Supply, 115 VAC, 1 each Each
      3,666.65
      Online Exclusive
      3,835.00
      Save 168.35 (4%)
      PSHV00A PowerEase™ Touch HV Power Supply Adapters Each
      110.00
      PSHV00 PowerEase™ Touch HV Power Supply, 115 VAC Each
      3,929.65
      Online Exclusive
      4,115.00
      Save 185.35 (5%)
      PS0120 PowerEase™ Touch 120W Power Supply, 115 VAC, 1 each Each
      951.65
      Online Exclusive
      994.00
      Save 42.35 (4%)
      PS0350 PowerEase™ Touch 350W Power Supply, 115 VAC, 1 each Each
      1,620.65
      Online Exclusive
      1,708.00
      Save 87.35 (5%)
      PowerEase Touch welcome packs
      For Research Use Only. Not for use in diagnostic procedures.