IHC Comparison of Click-iT® Plus TUNEL assay with competitor assay
Formalin-fixed paraffin-embedded (FFPE) mouse kidney was treated with DNase. After treatment, either the Click-iT® Plus TUNEL assay with Alexa Fluor® 594 dye (red) or the Roche In situ Cell Death TMR (orange) was used to detect the fragmented DNA. The cells were counterstained with Hoechst 33342 (blue). The GFP expressing epithelial cells are clearly visible (green).
Formalin-fixed paraffin-embedded (FFPE) mouse kidney was treated with DNase. After treatment, either the Click-iT® Plus TUNEL assay with Alexa Fluor® 594 dye (red) or the Roche In situ Cell Death TMR (orange) was used to detect the fragmented DNA.  The cells were counterstained with Hoechst 33342 (blue).  The GFP expressing epithelial cells are clearly visible (green).
Related Products
Catalog # Name Size Price (USD) Qty
C10617 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
849.65
Online Exclusive
878.00
Save 28.35 (3%)
C10618 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
836.65
Online Exclusive
864.00
Save 27.35 (3%)
C10619 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
839.65
Online Exclusive
868.00
Save 28.35 (3%)
Related Images