Neural somata in mouse cerebellum labeled with NeuroTrace® 530/615 red-fluorescent Nissl stain and DAPI.
Neural somata labeled with NeuroTrace® 530/615 red-fluorescent Nissl stain (Cat. no. N21482). Nuclei in this mouse cerebellum section were counterstained with DAPI (Cat. no. D1306, D3571, D21490). The image is a composite of two micrographs acquired using filters appropriate for tetramethylrhodamine and DAPI.
Neural somata labeled with NeuroTrace® 530/615 red-fluorescent Nissl stain (Cat. no. N21482).  Nuclei in this mouse cerebellum section were counterstained with DAPI (Cat. no. D1306, D3571, D21490). The image is a composite of two micrographs acquired using filters appropriate for tetramethylrhodamine and DAPI.
Related Products
Catalog # Name Size Price (USD) Qty
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D21490 DAPI, FluoroPure™ grade, 10 mg, 10 mg Each
225.65
Online Exclusive
231.00
Save 5.35 (2%)
D3571 DAPI (4',6-diamidino-2-phenylindole, dilactate), 10 mg, 10 mg Each
151.65
Online Exclusive
156.00
Save 4.35 (3%)
N21482 NeuroTrace™ 530/615 Red Fluorescent Nissl Stain - Solution in DMSO Each
296.00
Related Images