Build your flow cytometry panels for FREE with our scientists using our Panel Design Service or utilize our Panel Builder Tool to self-build your panels. We provide options for all experience levels when it comes to designing your flow cytometry panel.
We provide free flow cytometry panel design service for all customers
Getting started with our free panel design service
We have a team of technical support scientists available to help you. Our team provides this Panel Design Service for those who want help with building their panel every step of the way.
Request Panel Design with a Scientist
We provide free flow cytometry panel builder tool for self-panel design
Getting started with the flow cytometry panel builder tool
Choosing the optimal combination of fluorochromes can be simplified with a guided method. The Flow Panel Builder offers a curated and customizable approach to the panel building process.
Go to Panel Builder Tool
 Panel Design ServicePanel Builder Tool
WhatTake advantage of expert insight when:
  • Designing a new panel
  • Troubleshooting an existing panel
  • Revising and optimizing an existing panel with new markers
Intuitive interface that:
  • Organizes fluorochrome marker selections
  • Reports predicted spillover values
  • Shows fluorescence spectra according to your cytometer configuration
  • Offers a pre-configured panel match
Target usersAll flow cytometry users (all users—from naïve to experienced—are welcome to use)All flow cytometry users confident in their ability to design/optimize panels on their own
Time frameInitial response: within 24 hoursInstant
WhoLive scientists with panel design expertiseWebtool
Product purchaseNot needed
CostFREE (multiple-time usage, no catch)
What to expectPanel design, panel optimization, complete end-to-end service with final panel design; review with a minimum of 2 expertsDesign panels in 5 easy steps
  • Pre-configured panels—quick and easy access to panels designed by our specialists including many with supporting datasets
  • Streamlined input interface gets you to your panel more quickly
Mode of communicationFirst, complete the panel design service request form; our experts will then contact you by email/phone as neededSaved panels can still be used to communicate with the experts for “Free Panel Design Service”
Get your flow cytometry panels designed in 4 easy steps
Be the spark story - Berencia
Inspiring flow cytometry scientists, we create new multicolor panels at no cost for you.
Video Player is loading.
Current Time 0:00
Duration 3:16
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Getting started with the flow cytometry panel builder tool
    Choosing the optimal combination of fluorochromes can be simplified with a guided method. The Invitrogen Flow Panel Builder offers a customizable panel building process to fit your flow cytometry experimental needs, whatever your experience level.
    Video Player is loading.
    Current Time 0:00
    Duration 8:21
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 8:21
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Video: how to use the panel builder
      Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
      Video Player is loading.
      Current Time 0:00
      Duration 14:02
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 14:02
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Video: how to design spectral flow cytometry experiments
        Watch the video to create a panel for your spectral instrument
        List of available fluorophores based on their usage, benefits, and intended applications with Flurophore Selection Guide.
        See individual excitation and emission plots on the Invitrogen SpectraViewer.
        Tips and Tricks for Panel Design
        Video Player is loading.
        Current Time 0:00
        Duration 0:37
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 0:37
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Tip 1: Minimize spill-over by using the correct and separate channel
          Any time you have markers that you know will be co-expressed on your cells of interest, make sure to space them out into separate channels. If you will need to use any adjacent channels, that's where you would put any markers that are mutually exclusive so that they'll still be easy to distinguish.
          Video Player is loading.
          Current Time 0:00
          Duration 0:39
          Loaded: 0.00%
          Stream Type LIVE
          Remaining Time 0:39
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Tip 2: Intracellular targets need special buffer for fixation and permeabilization for staining
            You'll also want to keep in mind the buffer that you're using to fix and permeabilize your cells, as we have several options. When you're looking at cytoplasmic targets, what the buffer is appropriate may not be the same as when you're looking at nuclear targets, because you want to make sure that you still have access to your antigens without over-fixing your epitopes.
            Video Player is loading.
            Current Time 0:00
            Duration 0:39
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 0:39
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Tip 3: Viability dyes are required to find live cells
              A third tip I wanted to share with you is to always include a viability dye in your staining panel. This will help eliminate any false positives that are caused by dead cells or debris, because those can be sticky. You have a lot of options for choosing a viability dye, so you don't need to design your panel around them. You can build out the rest of your panel and optimize your core markers, and then fit in a viability dye in an empty channel.
              Video Player is loading.
              Current Time 0:00
              Duration 0:30
              Loaded: 0.00%
              Stream Type LIVE
              Remaining Time 0:30
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Tip 4: Save your bright fluorochromes for dim targets
                As you're building out your basic panel and you want to incorporate some more antigens, make sure you're keeping the density of your antigen expression in mind. So if you have antigens with low or unknown expression, those would be ones that you want to assign to your brightest dyes, such as PE or APC.
                Video Player is loading.
                Current Time 0:00
                Duration 0:40
                Loaded: 0.00%
                Stream Type LIVE
                Remaining Time 0:40
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  Tip 5: Try to combine negative markers in one channel (dump channel) to save space on your panel
                  A helpful trick when you want to exclude a lot of cell types at once without having to suck up multiple channels for that would be to use a dump channel. This is where you're placing all the antibodies that identify your cells that are not of interest into the same channel with the same fluorochrome, and then those can be easily gated out and all of the cells negative for the dump channel would be those that you use for your analysis going forward.
                  Immunology at work
                  Learn immune cell type markers and protocols. The Invitrogen Immunology at Work Resource Center is a learning center with technical content designed for new and experienced life scientists alike exploring the field of immunology.
                  Get started on your experiments
                  Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
                  Brilliant Violet and PE CF dyes are subject to proprietary rights of Becton, Dickinson and Company.
                  Cy™ is a trademark of Amersham Biosciences Corp. Cy dyes are subject to proprietary rights of Amersham Biosciences Corp and Carnegie Mellon University and are made and sold under license from Amersham Biosciences Corp only for research and in vitro diagnostic use.
                  BRILLIANT VIOLET™ is a trademark or registered trademark of Becton, Dickinson and Company or its affiliates, and is used under license. Powered by Sirigen™.