Food Allergens Testing Information
Identifying allergens in foods
Allergens pose a health risk, with up to 5% of the world’s population affected. Food allergen detection and quantification of proteins and protein residues is essential for safety and food labeling purposes.
LC-MS/MS using high-resolution accurate mass (HRAM) instrumentation is able to screen for whole proteins following tryptic digestion. This technique allows for the identification of multiple allergenic peptides in a food. The result can be that only one injection is required, making it an ideal tool for both qualitative and quantitative analysis.
Allergen testing is commonly carried out using ELISA and PCR techniques. In food manufacturing labs these techniques are quick, easy to use, and cost effective, and are used with a wide variety of foodstuffs. However, as developments have been made, it has been shown that certain proteins in processed foods such as eggs and milk can be difficult or even impossible to detect. Analytical techniques such as LC-MS/MS allow multiple allergens to be detected in a single analysis, with fewer false negatives, greater sensitivity, specificity, accuracy and reliability.
Join community
The most common food allergens are:
  • Milk
  • Eggs
  • Soybeans
  • Peanuts
  • Tree nuts
  • Wheat
  • Fish
  • Shellfish
Regulations
Allergens must be clearly stated by manufacturers on food labels, showing food source ingredients, additives and possible contamination of products where machinery is used for multiple processes. Failure to do so can lead to serious health and financial consequences for the industry.
There are various regulations throughout the world. Europe references EU regulation number 1169/2011 , which provides food information to consumers, requiring labeling and mandatory allergen information. The USA has The Food Allergen Labeling and Consumer Protection Act of 2004 . The act includes all foods apart from those regulated by other Federal agencies; these are poultry, most meats, certain egg products and the majority of alcoholic beverages.
Featured resources
Customer case study
Read about the challenges of identifying allergens in different matrices and how mass spectrometry is being used to detect them.
On-demand webinar
Learn about the essentials of food allergen analysis. Speakers from FARRP will discuss novel strategies for method development, which harness the power of HRAM-MS.
Featured videos
The Importance of Methods to Detect and Quantify Food Allergens
Dr. Melanie Downs from FARRP gives details on food allergies, including preventative controls and regulations and allergen analysis.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextThe Importance of Methods to Detect and Quantify Food Allergens
          Dr. Melanie Downs from FARRP gives details on food allergies, including preventative controls and regulations and allergen analysis.
        2. Now Playing
          Up NextNew Developments in Mass Spectrometry Allergen Methods
          Dr. Phil Johnson from FARRP gives details on the mass spectrometry methods he employs to detect allergens.
      Other allergy testing products
      As the world leader in in-vitro allergy diagnostics, we supply seven out of ten allergy laboratory tests worldwide. Thermo Scientific lmmunoCAP lab tests accurately measure specific lgE antibody blood levels, as well as other important allergy risk markers. ImmunoCAP is one of the most important tools for understanding allergy, helping clinicians make sense of symptoms such as rhinitis, eczema, wheezing and asthma.
      Learn more ›
      Food and Beverage Resource Library
      Access a targeted collection of scientific application notes, case studies, videos, webinars and white papers for food microbiology, manufacturing and processing, beverage testing, analytical testing, and authenticity information.