anti-frag-showcasw
We offer optimized reagents and kits based on immobilized papain, pepsin, and ficin proteases to selectively cleave IgG and IgM immunoglobulin molecules into Fab, Fab′, F(ab′)2 and Fc antibody fragments for use in immunodetection and immunoassay procedures.
Fab preparation kits
Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.
Catalog # Name Size Price (USD) Qty
44985 Pierce™ Fab Preparation Kit Each
848.00
44685 Pierce™ Fab Micro Preparation Kit Each
626.00
F(ab′)2 preparation kits
Selectively digest whole human and other IgG antibodies with immobilized pepsin protease to make F(ab′)2 fragments that retain antigen binding activity.
Catalog # Name Size Price (USD) Qty
44688 Pierce™ F(ab')2 Micro Preparation Kit Each
564.65
Online Exclusive
604.00
Save 39.35 (7%)
44988 Pierce™ F(ab')2 Preparation Kit Each
726.65
Online Exclusive
814.00
Save 87.35 (11%)
Production-FAbs-FAb2-fragments-intact-mouse-IgG1-Ficin
Production of Fab or F(ab′)2 fragments from intact mouse IgG1 using ficin
Mouse IgG1 Fab and F(ab′)2 preparation kits
Efficiently digest mouse IgG1 antibodies into Fab or F(ab′)2 fragments with immobilized ficin protease.
Bulk Quantities and Custom Products
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Complete request form
Biomarker quantitation assay guide
This 72-page guide provides detailed information about different tools for protein and RNA quantitation. Download this valuable technical resource that covers technologies useful for cancer and inflammation research, immunology, neurology and more. Learn more about how antibody pairs, ELISA kits, and multiplex kits for the Invitrogen Luminex platform may help advance your research.
Download the Biomarker quantitation assay guide
Biomarker quantitation assay guide cover
For Research Use Only. Not for use in diagnostic procedures.