Complete protein gel resupply packs—all you need to run 100 gels
Invitrogen Mini gel boxes, running buffer and protein ladder for gel electrophoresis
Protein gel resupply packs are a cost-effective solution that provide all the necessary components to run 100 protein gels. These packs combine trusted Invitrogen precast gels with precise quantities of running buffer and protein ladder, all conveniently bundled together for easy ordering.
Resupply packs contain:
  • 10 protein gel boxes
  • Running buffer
  • Prestained protein ladder
The products included in protein gel resupply packs have been carefully formulated and extensively tested to help ensure optimal compatibility and performance with Invitrogen precast gels. This meticulous attention to detail enables excellent protein separation and reliable results, helping enhance the overall quality of your experiments. Furthermore, protein gel resupply packs offer:
  • Extended shelf life—Bolt and NuPAGE Bis-Tris protein gels can be stored for up to 16 months at room temperature, allowing you to stock up and save without worrying about product degradation.
  • Protein Gel Performance Guarantee—These gels undergo rigorous quality testing to help ensure they meet exceptional standards of performance, reliability, reproducibility, and consistency.
    Read more about our Protein Gels Performance Guarantee
  • Midi WedgeWell format gels now available—high sample throughput meets high sample loading capacity. Run up to 26 samples per gel without compromising on sample loading volume.
Streamline your gel electrophoresis workflow to help reduce costs and have confidence in using premium products that are specifically designed to work seamlessly together. Upgrade your protein gel experiments today and experience the exceptional performance that Invitrogen is known for.
Bis-Tris Gels
Tris-Glycine Gels
Tris-Acetate Gels
Bis-Tris Gels
Tris-Glycine Gels
Catalog # Name Size Price (USD) Qty
STM5015-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 12%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM5012-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM5008-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM5014-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 12%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM4008-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, 1 Kit 1 kit
2,735.00
STM5011-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4011-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, 10 x 10 Gels kit 1 kit
2,735.00
STM5009-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4013-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, 10 x 10 Gels kit 1 kit
2,735.00
STM5013-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4009-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, 1 Kit 1 kit
2,735.00
STM4010-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, 10 x 10 Gels kit 1 kit
2,735.00
STM4012-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, 10 x 10 Gels kit 1 kit
2,735.00
STM5010-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
Tris-Acetate Gels
Bis-Tris Gels
Tris-Glycine Gels
Tris-Acetate Gels
Bis-Tris Gels
Tris-Glycine Gels
Catalog # Name Size Price (USD) Qty
STM5015-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 12%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM5012-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM5008-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM5014-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 12%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
STM4008-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, 1 Kit 1 kit
2,735.00
STM5011-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4011-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, 10 x 10 Gels kit 1 kit
2,735.00
STM5009-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4013-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, 10 x 10 Gels kit 1 kit
2,735.00
STM5013-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, WedgeWell™ format, 26-well, 1 Kit 1 kit
2,795.00
STM4009-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 20%, 1 Kit 1 kit
2,735.00
STM4010-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, 10 x 10 Gels kit 1 kit
2,735.00
STM4012-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 10%, 10 x 10 Gels kit 1 kit
2,735.00
STM5010-RS10 Novex™ Midi Tris-Glycine Plus Resupply Pack, 4 to 12%, WedgeWell™ format, 20-well, 1 Kit 1 kit
2,795.00
Tris-Acetate Gels
Need to add a gel electrophoresis tank to your gel electrophoresis essentials? See our Protein Gels Welcome Packs
Protein Gel Selection Guide
Protein Gel Selection Guide
Find an excellent mini or midi gel for your protein electrophoresis experiment.
Gel Selection Chart
Protein Gel Conversion Tool
Find an exceptional Invitrogen mini or midi gel to replace your current precast gel from another supplier.
Protein Gel Options Brochure
Gel Migration Chart
Download the Protein Gel Guide for a printable gel migration chart.
Still unsure which precast gel works best for your research needs? Download the Gel Selection Chart
Protein Gels Performance Guarantee
Protein Gels Performance Guarantee
We stand behind the performance of our protein gels.
Learn more
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.