Organism TypeHuman hamster hybrid cell line - transfected with two human genesThe 16-9 hamster human somatic cell hybrid line is the CHO-K1 hybrid containing a translocation of the long arm of human chromosome 6 encoding the IFN-R1 gene and a transfected human HLA-B7 gene (Soh et al., 1993).
TissuesOther
PhenotypeAdherent
Primaryno
ApplicationStable Cell Transfection
Products
Catalog # Name Size Price (USD) Qty
10131035 Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL), 20 mL Each
179.65
Online exclusive
197.00
Save 17.35 (9%)
18324012 Lipofectamine™ Transfection Reagent, 1 mL Each
650.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.