Search Thermo Fisher Scientific
Organism Type | Insect | Derived from Drosophila (fruit fly) embryo; doubling time is 20 to 24 hours. These cells are also known as S2 or SL2 cells. These cells can be grown in serum-supplemented or serum-free media in both adherent or suspension conditions. ATCC CRL-1963 |
Tissues | Embryo | |
Phenotype | Either | |
Primary | no | |
Application | Cell Culture / Growth Conditions,Nucleic Acid Purification,Protein Expression,RNAi,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||