Cell Therapy Systems (CTS)
細胞治療系統—臨床級細胞治療生產的可靠選擇
當您從基礎細胞治療研究過渡到臨床應用時,高品質的GMP級細胞治療原材料和充分的文件準備,是第一次就成功的重要關鍵。細胞治療系統(CTS)產品為臨床幹細胞治療及免疫療法的研究和生產,提供了經驗證的產品選擇,因此您可以放心地將細胞治療過渡到臨床應用。除了提供無血清、無異種和無動物來源的製劑外,我們GMP等級的CTS產品還具有針對細胞和基因療法的特定用途聲明、廣泛的安全性測試和充足的監管文件支持。我們採取這些嚴格措施來提供優質的原材料,以幫助客戶最大程度地降低風險、減輕品質管控系統的負擔,並滿足監管需求—讓CTS產品成為您從實驗室過渡到臨床的最佳選擇。
GMP生產規範
  • 原材料的生產符合醫材的cGMP、21 CFR Part 820 USP <1043> 標準和Ph Eur 5.2.12規定。
  • 我們的原材料生產基地已獲得FDA註冊,以及ISO 13485和ISO 9001認證。
測試與監管
  • 可追溯性文件—包括藥物主文件,和/或監管支持文件,以及原產地證書。
  • 廣泛的安全性測試—包括無菌、內毒素和黴漿菌。
經驗證且可靠的使用
  • 已應用於FDA批准的CAR-T療法和首個FDA批准的治療性癌症疫苗。
  • 已應用於200多項臨床試驗。
Gibco CTS Rotea逆流離心系統
Gibco CTS Rotea Counterflow Centrifugation System
這是一款小巧、靈活的封閉細胞處理系統,為了簡化並加速您的細胞療法開發而設計,從而實現:
  • 處理過程靈活—使用者可自行編程軟體,讓您可創建並優化各式各樣的細胞分離、洗滌和濃縮方案。
  • 優異的細胞回收率和活性—溫和的處理流程使細胞回收率>95%,同時保持細胞活性。
  • 低輸出體積—透過專利技術可輸出低至5 mL的濃縮液。
  • 實現從研究過渡到商業化生產—封閉式的一次性試劑盒可確保無菌處理,OPC-UA接口可連接至符合21 CFR Part 11的系統。
了解更多    申請儀器展示    洽詢價格
Gibco CTS Xenon電穿孔系統
Gibco-CTS-Xenon-Electroporation-Instrument
封閉式、可擴展的電穿孔系統,可用於符合GMP的細胞療法生產。
  • 處理量大且高速—在25分鐘以內,可轉染高達2.5 x 109 個T細胞/25 mL。
  • 規模可擴展,經過驗證且可靠的性能和細胞活力—使用CTS TrueCut Cas9蛋白可實現高達90 %的基因敲除和80 %的細胞活力。
  • 製程靈活性—使用者可編程系統,讓您從製程開發到商業化生產的過程中,都能根據各種細胞類型和有效承載,創建並優化電穿孔方案。
  • 高效的非病毒性轉染—可用於遞送DNA、RNA和蛋白質等有效承載。
  • 封閉式系統處理製程—Xenon MultiShot Electroporation Cartridge有助於實現與PVC或 C-Flex™ 管的無菌焊接。
了解更多    申請儀器展示    洽詢價格
Gibco CTS Xenon電穿孔系統
細胞治療應用
實現GMP等級的細胞治療開發
GMP設施和卓越生產
超過30年的GMP製造經驗使我們能夠提供高品質的GMP等級細胞治療原材料。我們的原材料生產基地已通過ISO 13485和ISO 9001認證,並獲得FDA註冊。我們策略規劃的生產設施,使我們能夠不間斷地向全球所有客戶提供最優質的產品。
優質生產嚴格符合法規要求
Gibco CTS產品是專為細胞治療應用而設計。製造原材料的方法和品管符合醫材GMP、21 CFR Part 820規定,並遵循USP <1043> 和Ph Eur 5.2.12指南。
安全測試
Gibco CTS培養基和試劑均為GMP等級,並經過嚴格的QC檢測,以確保其無菌性,以及是否存在內毒素和黴漿菌(僅對可適用產品)。
法規文件
詳盡的可追溯性文件,包括FDA藥物主文件和/或法規支持文件、分析證書和原產地證書,透過幫助支持您的監管申報並降低整個過程風險,從而減輕品質管制系統的負擔。
臨床和商業化應用
Gibco CTS產品已在全球200多項臨床試驗中,應用於細胞療法的商業化生產。我們致力於透過確保商業使用權,來支持客戶的GMP等級細胞治療生產製造。
專業支持
由經驗豐富的細胞治療專家和區域技術部門,組成資深的法規支援團隊。
Gibco CTS紀錄短片系列
Episode 1: Opportunities in cell therapy translational research
Representatives from ViaCyte, Inc and City of Hope talk about the opportunities and excitement surrounding the potential for translating immunotherapy and regenerative medicine studies to the clinic.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextEpisode 1: Opportunities in cell therapy translational research
          Representatives from ViaCyte, Inc and City of Hope talk about the opportunities and excitement surrounding the potential for translating immunotherapy and regenerative medicine studies to the clinic.
        2. Now Playing
          Up NextEpisode 2: Challenges in cell therapy translational research
          Challenges facing the cell therapy industry include supply chain, scale up, quality controls, and manufacturing process development. Hear comments from representatives from ViaCyte, Inc and City of Hope.
        3. Now Playing
          Up NextEpisode 3: Impact of raw materials on cell therapy development
          Raw material selection is key for cell therapy manufacturing endeavors, as it can impact cost, process, quality, and reproducibility. Hear comments on this topic from representatives from ViaCyte, Inc and City of Hope.
        4. Now Playing
          Up NextEpisode 4: Improving cell therapy systems with raw material selection
          The use of agents that can exacerbate potential immune response to cell therapies is a critical consideration for manufacturers. Hear thoughts from representatives from ViaCyte, Inc and City of Hope on the use of xeno-free reagents.
        5. Now Playing
          Up NextEpisode 5: Importance of scalable cell therapy manufacturing processes
          Representatives from ViaCyte, Inc and City of Hope talk about key considerations when moving a process from lab scale to commercial scale, including individualized cell requirements, supply chain, and manufacturing needs.
        6. Now Playing
          Up NextEpisode 6: Benefits of documentation in cell therapy development
          Components utilized in cell therapy manufacturing processes need to be accompanied by the reliable safety documentation. Hear comments on this topic from representatives from ViaCyte, Inc and City of Hope.
        7. Now Playing
          Up NextEpisode 7: Choosing the ideal cell therapy product supplier
          Strong quality management system, close collaborative relationship, and superior product support, are critical in choosing a supplier. Listen to representatives from ViaCyte, Inc and City of Hope share their thoughts on this topic.
      leaf icon 減少浪費
      盡最大努力減少實驗室中的浪費,每一點累積都有所幫助。與目前市面上同類培養基瓶相比,1 L Gibco瓶裝使用的原材料減少了38 %(0.5 L瓶裝減少了32%)。這將降低不可再生資源的消耗,有助於減少您的碳足跡。
      深入了解有關可持續解決方案
      leaf icon 永續包裝
      永續包裝必須保護內部產品—但這並不意味著必須犧牲永續性。GlutaMAX-I培養基添加劑可在常溫下運輸,每年這些產品包裝和運輸的碳足跡減少了近103噸(二氧化碳當量)。
      深入了解有關可持續解決方案
      Resources
      支持
      本頁面提及產品的預期用途有所相同。有關特定的預期用途聲明,請參閱產品標籤。