CTS™ Immune Cell Serum Replacement (SR)
TrueCut Cas9 is now available as a CTS-prototype for preclinical and translational research. Learn more »
CTS™ Immune Cell Serum Replacement (SR)
Gibco™

CTS™ Immune Cell Serum Replacement (SR)

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells when added as a serum substitute to supplement a basal cell culture medium such as Gibco CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium.
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzleLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A2596101Promo Image50 mL
A4702901Promo Image250 mL
A2596102Promo Image500 mL
A4702902Promo Image1 L
Catalog number A2596101
Price (USD)
259.00
Each
In stock
Add to cart
Quantity:
50 mL
Recurring order eligible. Learn more »
Price (USD)
259.00
Each
Add to cart
Ask our AI about this Product
CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells when added as a serum substitute to supplement a basal cell culture medium such as Gibco CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium.

Serum replacement

Human serum, a key component in many immune cell culture protocols, is expensive, inherently variable, and requires extensive safety testing prior to use when manufacturing a cGMP-compliant adoptive cell therapy product. CTS Immune Cell SR was developed to provide a serum-free, xeno-free complete medium that would minimize the supply and safety risks associated with human serum.

This defined and robust formulation can replace supplementation with human serum in several different base media without sacrificing any performance of your cells. CTS Immune Cell SR eliminates the time and cost of qualifying human serum, enabling a seamLess transition from research to clinical applications.

Immune cell qualified

CTS Immune Cell SR supports equivalent or improved results for multiple immune cell types, when used at similar concentration as human serum.

Closed workflow compatible

CTS Immune Cell SR is available in bioprocess container format. The chamber is constructed from our highest quality Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock or MPC quick connect provides versatile connection options for integration into your closed process.

CTS quality

Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing

Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.

*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeHuman Immune Cells
For Use With (Application)Cell and Gene Therapy Research, Development and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeSerum Replacement
Quantity50 mL
Shelf Life18 months from date of manufacture
Shipping ConditionDry Ice
ClassificationXeno-free
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Storage Conditions: -5°C to -20°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3155380Certificate of AnalysisJun 13, 2025A2596101, A2596102
3155376Certificate of AnalysisJun 08, 2025A2596101, A2596102
3155374Certificate of AnalysisMay 25, 2025A2596101, A2596102
3135578Certificate of AnalysisMay 22, 2025A4702901, A4702902
3013490Certificate of AnalysisMay 08, 2025A4702901, A4702902
5 results displayed, search above for a specific certificate

Safety Data Sheets

Limited Use Label Licenses (LULL)

'

Notice to Purchaser: Resale of this product is expressly prohibited. No right to resell this product or any of its components is conveyed expressly, by implication, or by estoppel. For information on obtaining additional rights, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.

'

Frequently asked questions (FAQs)

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

After completely thawing the initial product, it can be aliquoted and refrozen for another usage. The thawed aliquots are stable for 4 weeks when stored at 2-8 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
A Rapid Cell Expansion Process for Production of Engineered Autologous CAR-T Cell Therapies.
Authors:Lu TL, Pugach O, Somerville R, Rosenberg SA, Kochendefer JN, Better M, Feldman SA
Journal:Hum Gene Ther Methods
PubMed ID:27897048
The treatment of B-cell malignancies by adoptive cell transfer (ACT) of anti-CD19 chimeric antigen receptor T cells (CD19 CAR-T) has proven to be a highly successful therapeutic modality in several clinical trials.(1-6) The anti-CD19 CAR-T cell production method used to support initial trials relied on numerous manual, open process steps, ... More
1 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection