EVOS S1000 Spatial Imaging System for Multiplexed Tissue Imaging
The EVOS S1000 Spatial Imaging System streamlines the process of tissue imaging for spatial localization of proteins
The Invitrogen EVOS S1000 Spatial Imaging System is a widefield spectral imaging system that combines multiplex spectral fluorescence, transmitted brightfield, and color brightfield detection. Coupled with EVOS spatial imaging software, it provides seamless image processing, single-round spectral unmixing of up to 8-plex (+ nuclei detection), and powerful image visualization, all within a user-friendly platform. The spectral unmixing software enables the EVOS S1000 Spatial Imaging System to image a single round of 9-plex multiplex immunofluorescence faster than instruments utilizing cyclic technology. The EVOS S1000 Spatial Imaging System is excellent for scientists entering the spatial biology field, including those specializing in cancer biology, histology, immunology, and neurobiology who require highly resolved images. 
Video Player is loading.
Current Time 0:00
Duration 1:11
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:11
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    Customer story:  SciLifeLab, Stockholm, Sweden, Spatial Proteomics Unit
    Using the Invitrogen EVOS S1000 Spatial Imaging System simplifies the tissue imaging process and provides multiplex data in less than one hour.
    Features
    • Fast—Scan, spectrally unmix, and stitch 9-plex image of a 1 cm2 tissue area at 10X in about 20 min
    • Versatile—Use of many types of fluorophores, antibodies, RNA probes, and chromagens from most vendors
    • Comprehensive—Perform spectral fluorescence, transmitted brightfield, phase-contrast, and color brightfield imaging
    • Sensitive—Detect intricate multiplexed staining patterns within tissues with a 4.2 megapixel, 16-bit sCMOS camera
    • Variable magnification—Easily use 2.5x–40x objectives available for the 5-position slider
    • Efficiency—Streamline image acquisition with automated laser and software-based autofocus routines
    • Simple—Intuitive software makes image collection, navigation, and data retrieval simple for users of all experience levels
    • Flexible—TIFF, OME-TIFF, Pyramidal structure available so you can use the data analysis software of your choice
    • Highly resolved data—Our spectral solution reduces fluorescence crosstalk and produces highly resolved images
    3-panel microscopic tissue image
    Figure 1. Stained tonsil tissue captured using the EVOS S1000 Spatial Imaging System: a transmitted brightfield image in black and white, a color brightfield image showcasing purple H&E staining, and a multicolor spectral fluorescence image. EVOS S1000 Spatial Imaging System is capable of different modalities.
    Video Player is loading.
    Current Time 0:00
    Duration 25:31
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 25:31
    1x
      • Chapters
      • descriptions off, selected
      • subtitles off, selected
      • en (Main), selected
      Advancing multiplex imaging for spatial proteomics research
      Watch and learn about the advancements in multiplex imaging for spatial protein research. Discover how the Invitrogen EVOS S1000 Spatial Imaging System simplifies workflows, supports multiple dyes, and enables fast, high-resolution imaging. Learn about the Invitrogen Aluora Spatial Amplification Reagents and how they provide high sensitivity and bright signals for flexible antibody usage. Understand how fast and simple multiplexing antibodies can be with primary antibodies conjugated to fluorophores and when to use conjugation kits including the Invitrogen ReadyLabel antibody conjugation kits.
      "With the new EVOS S1000, we can run more projects in parallel. It allows us to achieve multiplexing without needing to remove antibodies or fluorophores and it significantly reducing sample processing time"
      - Carolina Osés Sepúlveda, SciLife Stockholm, Sweden
      “The EVOS S1000 has a very fast software, I like the live [periscope] mode. I also like the simplicity of the unmixing workflow. It is really compatible with analysis software like QuPath!”
      -Giulia Bergamaschi, Amsterdam Universitair Medische Centra (UMC), The Netherlands
      "The software is very intuitive and easy to explain. The images are super crisp and it's great to be able to see the scanning progress in real time. The spectral unmixing algorithm really makes signals stand out”
      -Maria Lung, SciLife Stockholm, Sweden
      Overcome the limitations of traditional immunohistochemistry with the EVOS S1000 Spatial Imaging System and gain more information from your tissues
      The EVOS S1000 Spatial Imaging System transforms immunohistochemistry (IHC) by addressing the limitations of single- or double-staining methods, which are often overly simplistic, lose contextual data, and fail to identify complex cell types. Single-stained sections offer limited detail, even when compared (Figure 2) side-by-side. By enabling advanced multiplexing capabilities, the EVOS S1000 system allows for the colocalization of multiple signals within a single, stained tissue sample. This innovative approach in spatial biology helps EVOS S1000 Spatial Imaging System deliver a comprehensive snapshot of tissue microenvironments and architecture in their native state (Figure 3).
      Simplified tissue imaging workflow
      Unlike existing technologies that can take several days to weeks, the EVOS S1000 instrument can complete the imaging process within several hours, with the greatest time savings found in its capability to generate a fully stitched, unmixed, multiplexed image for multiple samples. With the EVOS S1000 Spatial Imaging System, you have the flexibility to use a variety of labeling technologies, allowing you to select and utilize your preferred antibodies and reagents. The instrument outputs OME-TIFF files that are compatible with any third-party analysis software such as Halo and QuPath (QuPath: Open source software for digial pathology image analysis. Scientific Reports (2017).)
      Workflow graphic
      Figure 10: The spatial biology multiplex workflow incorporates the EVOS S1000 Spatial Imaging System that multiplexes more biomarkers with the use of spectral unmixing.
      To build a panel for the EVOS S1000 Spatial Imaging system, start by choosing your targets, ensuring they align with your research goals. Next, create the panel by carefully picking out antibodies; use the Invitrogen SpectraViewer to determine where to place the 8 fluorophores labeling the antibodies plus DAPI. For the staining and labeling technology, use our Spatial Biology Reagent Selection Tool to chose what is right for your experiment. Finally, proceed with the imaging step to capture and analyze your samples.
      Benefits of spectral unmixing technology
      Perform imaging faster
      Accelerate your experiments with the EVOS S1000 Spatial Imaging System than traditional cyclic technologies. Complete a full round of 9-plex multiplex immunofluorescence faster using advanced software with spectral unmixing.
      bar chart of imaging time
      Reduce the chance for tissue damage
      Prevent extensive tissue damage. Iterative staining using bleaching or antibody-removal methods extends experimental time due to lengthy antibody and reagent incubations, and poses risks of epitope loss, tissue degradation, and incomplete fluorophore inactivation across multiple cycles.
      Illustration of imaging cycle advantages
      Find confidence in your experiments
      Gain peace of mind with the EVOS S1000 Spatial Imaging System automated unmixing matrix generation for imaging-stained multiplex tissue samples. The EVOS S1000 software automatically generates and applies an unmixing matrix to spectrally mixed multiplex tissue samples, clearly identifying fluorophore emissions that bleed into neighboring channels.
      software screenshot
      EVOS S1000 system–compatible antibodies and reagents
      Conjugated primary antibodies
      Detect numerous targets simultaneously using conjugated primary antibodies for multiplexing experiments. These antibodies have been tested on tissue samples and are available in formats compatible with the EVOS S1000 Spatial Imaging System and other microscopy platforms.
      Aluora Spatial Amplification Kits
      Detect up to 8 targets simultaneously in one tissue sample with Aluora Spatial Amplification Kits. These kits combine bright Aluora dyes with enzyme-mediated signal amplification to help overcome autofluorescence in the detection of rare targets.
      ReadyLabel Antibody Labeling Kits
      Overcome the challenges of assembling multiplex imaging panels by directly conjugating your primary antibody with dye colors optimized for spatial analysis.
      ViewRNA ISH Tissue Assay
      Detect multiple RNA targets within one tissue sample. The kit provides exceptional sensitivity and allows single-molecule detection of up to four RNA targets simultaneously.
      Ordering information
      Resources and support
      Spatial imaging protocol handbook
      Read and use trusted protocols for many types of imaging including 3D staining, multiplexing ISH in tissue, and other spatial biology protocols.
      Spatial biology resource center
      Find resources including webinars, conference posters, protocols, guides, handbooks, and publications to start your multiplex experiments.
      SpectraViewer
      Explore the excitation and emission spectra of fluorophores.
      Spatial Reagent Finder
      The Spatial Biology Reagents Selection Tool helps you find the reagents compatible with your multiplex IHC experimental needs.
      Application notes
      Scientific posters
      Selection guides
      Specification sheet
      Other articles
      How-to tutorial videos
      The Invitrogen EVOS S1000 Spatial Imaging System is only available in participating countries and regions including US, Canada, and Europe in 2024.
      HALO is a trademark of Indica Labs, Inc.
      For Research Use Only. Not for use in diagnostic procedures.