A variety of flask sizes filled with phenol-red medium
T225 flask volume and surface area
Start your scale-up with a T225 flask that has the surface and ancillary options you're looking for in a tissue culture flask. Choose a cell culture flask from a collection that includes straight-neck or angled-neck options and surface treatment alternatives that help ensure your healthy tissue cells expand and grow in the flask of your choice.
Some useful numbers such as surface area and volumes of dissociation solutions are given below for T225 flasks.
Please note: The number of cells on a flask will vary with cell type. For this table, 6.3 x 106 HeLa cells were seeded and the confluent flask contained 30 x 106 cells.
T225 flasks have a surface area of 225 cm2
Dissociation reagentVolume of reagentVolume of growth medium
TrypLE Express enzyme22.0 mL45–68 mL
TrypLE Select (1x) enzyme22.0 mL
TrypLE Select (10x) enzyme2.2 mL diluted to 22.0mL in DPBS/1 mM EDTA
Versene reagent22.0 mL
Trypsin22.0 mL
Sign up to receive our informative publications featuring blogs, webinars, and more as soon as they're available
A single T225 EasYFlask with angled neck
T225 flask: EasYFlask products
Ergonomically designed for maximal access to the growth surface, this low-profile flask optimizes incubator space and features angled walls for complete viewing under microscopes. Additional features include multiple cap options, and sterilization by gamma irradiation with non-pyrogenic certification.
EasYFlask product features
A BioLite surface treated flask with blue cap
Thermo Scientific BioLite T225 flask surface
Thermo Scientific BioLite cell culture treated flasks with filter cap and seal cap options suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
Caps come in both vented and plug seal varieties.
Catalog # Name Size Price (USD) Qty
130330 BioLite™ T225 Flask, Cell Culture-treated surface, Filter Cap Case of 25
255.65
Online Exclusive
262.00
Save 6.35 (2%)
130332 BioLite™ T225 Flask, Cell Culture-treated surface, Seal Cap Case of 25
252.65
Online Exclusive
260.00
Save 7.35 (3%)
Nunclon Delta treated T225 flasks
Composed of cell culture-treated polystyrene and surface treated with proprietary Nunclon Delta surface, these T225 flasks support optimal growth and attachment of cells to the growth surface.
Filtered caps allow for constant airflow while vent/close caps allow for sufficient gas exchange.
Explore cell performance with Nunclon Delta surfaces
Catalog # Name Size Price (USD) Qty
159934 T225 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 30
328.65
Online Exclusive
393.00
Save 64.35 (16%)
159933 T225 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 30
288.65
Online Exclusive
405.00
Save 116.35 (29%)
Related flask and cell culture resources
Surface selection: Achieving optimal cell growth and harvesting
Get an overview of technologies used to modify cultureware surfaces to help you select the right surface for your application.
  • Surface modifications to improve cell adhesion
  • Characterization of Nunc cell culture surfaces by adhesion properties
  • Alternative methods for advanced cell harvesting
Watch now
Establishing a dynamic micro-gravity spheroid 3D culture system
Learn about how to build specific 3D cell spheroid models and platforms to answer the pressing and relevant questions in cancer research.
  • Discerning the advantages and disadvantages of 2D versus 3D cell culturing approaches
  • Understanding the method of rotating micro-gravity bioreactors as a means to produce long-term spheroid cultures
  • Implementing spheroid cultures in cancer research
Watch now
cell-culture-cafe-230x195
Watch more cell culture webinars
Register for our next on-demand webinar event and sign up to receive everything we have on cell culture.
Visit the café
Featured article
Cell adhesion and growth on coated or modified glass or plastic surfaces
Growth substrates affect the adhesion, growth, morphology and differentiation of various cell types. However, the response to a given surface is cell type specific. Selection of a growth surface for cell culture should be based on cell culture performance and on application. In this technical bulletin, the growth of various cell types on plastic, soda lime glass and borosilicate coverglass was examined with surfaces that were either unmodified, coated with polylysine, or stably surface modified with non-biological reagents or electrical discharge. 
Download bulletin
Are you on the front lines of cancer research?
Video Player is loading.
Current Time 0:00
Duration 1:34
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:34
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Surface selection: Achieving optimal cell growth and harvesting
    Get an overview of technologies used to modify cultureware surfaces to help you select the right surface for your application.
    • Surface modifications to improve cell adhesion
    • Characterization of Nunc cell culture surfaces by adhesion properties
    • Alternative methods for advanced cell harvesting
    Watch now
    Establishing a dynamic micro-gravity spheroid 3D culture system
    Learn about how to build specific 3D cell spheroid models and platforms to answer the pressing and relevant questions in cancer research.
    • Discerning the advantages and disadvantages of 2D versus 3D cell culturing approaches
    • Understanding the method of rotating micro-gravity bioreactors as a means to produce long-term spheroid cultures
    • Implementing spheroid cultures in cancer research
    Watch now
    cell-culture-cafe-230x195
    Watch more cell culture webinars
    Register for our next on-demand webinar event and sign up to receive everything we have on cell culture.
    Visit the café
    Featured article
    Cell adhesion and growth on coated or modified glass or plastic surfaces
    Growth substrates affect the adhesion, growth, morphology and differentiation of various cell types. However, the response to a given surface is cell type specific. Selection of a growth surface for cell culture should be based on cell culture performance and on application. In this technical bulletin, the growth of various cell types on plastic, soda lime glass and borosilicate coverglass was examined with surfaces that were either unmodified, coated with polylysine, or stably surface modified with non-biological reagents or electrical discharge. 
    Download bulletin
    Are you on the front lines of cancer research?
    Video Player is loading.
    Current Time 0:00
    Duration 1:34
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 1:34