IPTG (isopropylthio-β-galactoside) is an inducer of β-galactosidase activity in bacteria and is suitable for use with X-gal or bluo-gal to detect lac gene activity during cloning. Life Technologies offers IPTG in several sizes for added convenience.
Ordering Information
Catalog # Name Size Price (USD) Qty
15529019 IPTG Each
92.65
Online Exclusive
98.00
Save 5.35 (5%)
AM9462 IPTG (Dioxane-Free), 2 g Each
145.65
Online Exclusive
152.00
Save 6.35 (4%)
AM9464 IPTG (Dioxane-Free), 5 g Each
279.65
Online Exclusive
297.00
Save 17.35 (6%)