When working with RNA, it is important to protect the integrity and quality of RNA by eliminating RNase contamination. RNases—or ribonucleases—are enzymes that target and degrade RNA. Lab detergents and cleaning solutions are available to help keep lab equipment and surfaces RNase-free.
RNase decontamination via RNase inactivation
One way to combat RNase contamination is via RNase inactivation. This means treating the surfaces you suspect need RNase decontamination with solutions that contain chemicals known to be active against RNases.
Another common alternative to RNase decontamination is by autoclaving equipment, such as glassware and metalware. The baking procedure typically involves incubation in an oven at 450°F for two hours or more.
Read about RNase detection
RNase decontamination solutions
Treatment with RNase decontamination solutions is a useful way to eliminate RNases from equipment or lab surfaces. Thermo Fisher Scientific offers several products in the RNaseZap family—RNaseZap Solution, RNaseZap Wipes, and ElectroZap Solution—that collectively provide a comprehensive approach to help ensure that work surfaces, pipettors, and equipment are RNase-free.
  • Better RNase A decontamination than competitive products
Formats for RNase decontamination solutions include detergents and wipes.
RNase decontamination solutions in a detergent format
RNaseZap RNase Decontamination Solution product photo of the bottle and sprayer cap
RNaseZap RNase Decontamination Solution is a decontamination solution that inactivates and removes RNases. RNaseZap solution comes in a sprayer bottle so you simply spray the solution on contaminated surfaces and rinse off with RNase-free water. Main features of this solution include:
  • Glass, plastic, and surface decontaminant
  • Minimizes RNase contamination
ElectroZap RNase Decontamination Solution product photo of the bottle and sprayer cap
ElectroZap Electrode Decontamination Solution is an RNase decontamination solution that inactivates and removes RNases. This solution is formulated to decontaminate RNases on electrode surfaces. Main features of ElectroZap solution include:
  • Non-corrosive cleaning solution
  • Electrode surface decontaminant
  • Minimizes RNase contamination
Ordering information for RNase decontamination detergents
Catalog # Name Size Price (USD) Qty
AM9780 RNaseZap™ RNase Decontamination Solution, 250 mL Each
105.65
Online Exclusive
111.00
Save 5.35 (5%)
AM9782 RNaseZap™ RNase Decontamination Solution, 6 x 250 mL Each
469.65
Online Exclusive
497.00
Save 27.35 (6%)
AM9784 RNaseZap™ RNase Decontamination Solution, 4 L Each
919.65
Online Exclusive
962.00
Save 42.35 (4%)
AM9785 ElectroZap™ Electrode Decontamination Solution Each
101.00
RNase decontamination solutions in a towelette format
RNaseZap RNase Decontamination Wipes product photo of the container and packaging
RNaseZap RNase Decontamination Wipes contain the same RNaseZap solution in a convenient towelette form. The RNase decontamination solution contains ingredients known to inactivate and eliminate RNases. Main features of RNaseZap wipes include:
  • Glass and plastic surface decontaminant in wipes
  • Minimizes RNase contamination
Ordering information for RNase decontamination wipes
Catalog # Name Size Price (USD) Qty
AM9786 RNaseZap™ RNase Decontamination Wipes, 100 Sheets Each
122.65
Online Exclusive
128.00
Save 5.35 (4%)
AM9788 RNaseZap™ RNase Decontamination Wipes Refill, 300 Sheets Each
279.65
Online Exclusive
299.00
Save 19.35 (6%)
RNase decontamination solutions in a detergent format
RNaseZap RNase Decontamination Solution product photo of the bottle and sprayer cap
RNaseZap RNase Decontamination Solution is a decontamination solution that inactivates and removes RNases. RNaseZap solution comes in a sprayer bottle so you simply spray the solution on contaminated surfaces and rinse off with RNase-free water. Main features of this solution include:
  • Glass, plastic, and surface decontaminant
  • Minimizes RNase contamination
ElectroZap RNase Decontamination Solution product photo of the bottle and sprayer cap
ElectroZap Electrode Decontamination Solution is an RNase decontamination solution that inactivates and removes RNases. This solution is formulated to decontaminate RNases on electrode surfaces. Main features of ElectroZap solution include:
  • Non-corrosive cleaning solution
  • Electrode surface decontaminant
  • Minimizes RNase contamination
Ordering information for RNase decontamination detergents
Catalog # Name Size Price (USD) Qty
AM9780 RNaseZap™ RNase Decontamination Solution, 250 mL Each
105.65
Online Exclusive
111.00
Save 5.35 (5%)
AM9782 RNaseZap™ RNase Decontamination Solution, 6 x 250 mL Each
469.65
Online Exclusive
497.00
Save 27.35 (6%)
AM9784 RNaseZap™ RNase Decontamination Solution, 4 L Each
919.65
Online Exclusive
962.00
Save 42.35 (4%)
AM9785 ElectroZap™ Electrode Decontamination Solution Each
101.00
RNase decontamination solutions in a towelette format
RNaseZap RNase Decontamination Wipes product photo of the container and packaging
RNaseZap RNase Decontamination Wipes contain the same RNaseZap solution in a convenient towelette form. The RNase decontamination solution contains ingredients known to inactivate and eliminate RNases. Main features of RNaseZap wipes include:
  • Glass and plastic surface decontaminant in wipes
  • Minimizes RNase contamination
Ordering information for RNase decontamination wipes
Catalog # Name Size Price (USD) Qty
AM9786 RNaseZap™ RNase Decontamination Wipes, 100 Sheets Each
122.65
Online Exclusive
128.00
Save 5.35 (4%)
AM9788 RNaseZap™ RNase Decontamination Wipes Refill, 300 Sheets Each
279.65
Online Exclusive
299.00
Save 19.35 (6%)
TechNotes articles
Support
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments