Centifuges for almost any application
centrifuges applications 1200
Centrifuges for cell culture workflows
Centrifuges play a critical role in cell culture applications through various processes that involve the separation, concentration, and purification of cells. Here are some key ways in which centrifuges are utilized in cell culture:
  1. Cell Isolation and Separation: Centrifuges are used to isolate specific cell types from a heterogeneous mixture. By applying centrifugal forces, cells can be separated based on their size, shape, and density. This is particularly useful for isolating stem cells, immune cells, or other specific cell populations from blood or tissue samples.
  2. Cell Concentration: After isolation, the desired cells often need to be concentrated to achieve the necessary cell dose for therapeutic applications. Centrifugation can concentrate cells by removing excess fluids and unwanted cell types, ensuring that the final cell product is at the required concentration for effective therapy.
  3. Purification: Centrifuges help in the purification of cells by removing contaminants such as dead cells, cell debris, and other unwanted particles. This step is crucial to ensure that the cell product is pure and safe for therapeutic use.
  4. Cell Washing: Cells are often washed to remove any residual media, reagents, or contaminants before being used in therapy. Centrifugation allows for efficient washing by pelleting the cells and replacing the supernatant with a fresh buffer or medium.
  5. Density Gradient Centrifugation: This technique uses a density gradient medium to separate cells based on their buoyant density. It is commonly used to isolate specific cell types or subpopulations, such as different types of immune cells, from a mixed cell population.
  6. Exosome Isolation: Exosomes are small vesicles secreted by cells that have potential therapeutic applications. Ultracentrifugation is often employed to isolate exosomes from cell culture supernatants or body fluids, enabling their use in cell-free therapies.
  7. Quality Control: Centrifugation is also used in quality control processes to assess the viability, purity, and functionality of cell products. By separating and analyzing cell populations, researchers can ensure that the cell therapy product meets the required standards.
Cell and gene therapy
On-demand webinar: Role of batch centrifugation during cell therapy manufacturing
cgt batch cent webinar
Application note: Downstream processing of primary T cells 
cgt downstream tcells
Application note: Production of therapeutic plasmid DNA in single use fermentors
cgt plasma dna
Bioprocessing
Smart note: How can I maintain sterility while spending less time on cell culture harvest steps?
bio culture harvest
Smart note: How do you maximize cell pellet recovery and minimize freezer storage space in cell cultures?
bio cell pellet recovery
Exosome isolation and purification
White paper: High yield, high purity sEV purification scale-up method by zonal density gradient ultracentrifugation  
evox exosome whitepaper
Blog post: Unveiling Exosome Isolation and Purification Protocols: A Deep Dive into Ultracentrifugation Techniques
exosome blog
White paper: Exosome isolation from cell culture by ultracentrifugation: optimized protocols 
isolation exosome whitepaper
White paper: Isolation and purification of exosomes from blood plasma by ultracentrifugation using a two-step approach 
blood plasma exosome whitepaper
Application note: Exosome isolation with LYNX Superspeed Centrifuges
lynx exosome appnote
Clinical applications where centrifugation is important
  1. Blood Sample Processing: One of the most common clinical uses of centrifuges is the separation of blood components. By spinning blood samples at high speeds, centrifuges can separate plasma, serum, red blood cells, white blood cells, and platelets. This is essential for various diagnostic tests, including blood chemistry analysis, hematology, and serology.
  2. Urine Sample Processing: Centrifuges are used to process urine samples by separating solid particles (sediments) from the liquid. The sediments can then be examined microscopically for the presence of cells, bacteria, crystals, and other substances, aiding in the diagnosis of urinary tract infections, kidney disorders, and other conditions.
  3. Preparation of Platelet-Rich Plasma (PRP): PRP therapy involves using a patient's own platelets to promote healing of injured tissues. Centrifuges are used to concentrate platelets from the patient's blood, creating a PRP preparation that can be injected into the affected area to accelerate healing.
  4. Cell and Tissue Culture: Centrifuges are used to harvest cells from culture media, wash cells, and prepare cell pellets for further analysis or therapeutic use. This is particularly important in regenerative medicine and cell therapy applications.
  5. Molecular Biology and Genomics: In clinical research and diagnostics, centrifuges are used to extract and purify nucleic acids (DNA and RNA) and proteins from biological samples. This is critical for techniques such as polymerase chain reaction (PCR), sequencing, and various forms of electrophoresis.
  6. Microbiology: Centrifuges are used to concentrate microorganisms from clinical samples, such as sputum, urine, or blood, to facilitate their identification and susceptibility testing. This is important for diagnosing infections and determining appropriate antimicrobial treatments.
  7. Cytology: In cytology laboratories, centrifugation is used to prepare cell samples for microscopic examination. For example, in Pap smear tests, centrifuges help concentrate and prepare cervical cells for screening of precancerous and cancerous lesions.
  8. Immunoassays: Centrifugation is often a step in immunoassay protocols, where it is used to separate bound from free antigen-antibody complexes. This is crucial in various diagnostic tests, including enzyme-linked immunosorbent assays (ELISAs) and radioimmunoassays (RIAs).
  9. Cryopreservation: Before cryopreservation, cells and tissues are often centrifuged to concentrate and wash them, ensuring they are in the optimal state for freezing and long-term storage. This is particularly important for preserving stem cells, sperm, and other biological materials.
  10. Pharmaceutical Preparations: In the preparation of certain pharmaceutical products, centrifuges are used to purify and concentrate biologics, such as vaccines, monoclonal antibodies, and other therapeutic proteins.
Molecular biology
Technical note: Isolation of Plasma Membrane from Endothelial Cells Using the Thermo Scientific Sorvall MTX Micro-Ultracentrifuge and S55-A2 Rotor
CENT molbio plama technote
Application note: Traditional Methods for CsCl Isolation of Plasmid DNA by Ultracentrifugation
CENT molbio app note
Application note: DNA Preparation from Large Volume of Whole Blood using Thermo Scientific General Purpose Centrifuge with Fiberlite Rotors
CENT molbio gp rotors app note
Clinical diagnostics/ research
Application note: A comprehensive protocol for lipemia removal from serum using an ultra microcentrifuge
CENT clinical medifuge app note
Application note: Using Medifuge Centrifuge for Coagulation Testing through preparation of Platelet-Poor Plasma (PPP) 
CENT clinical medifuge app note
Application note: Using Thermo Scientific Small Benchtop Centrifuges for the Effective and Efficient Processing of Clinical Samples 
CENT clinical benchtop app note
Application note: Technical guide on efficient sample preparation for clinical diagnosis
CENT clinical sample prep app note
Blood banking
Application note: Blood banking applications using the Thermo Scientific Sorvall BP 8 and 16 and Cryofuge 8 and 16 Centrifuges
CENT blood cryofuge app note
Smart note: How can blood banks eliminate potential variables to ensure consistent product yield and reproducibility?
CENT blood banks smart note
Smart note: How can blood banking and bioprocessing production facilities enhance ergonomics, while simplifying operation for greater productivity during their separations?
CENT blood ergonomics smart note
Other
Application note: Isolating environmental DNA in the field using Thermo Scientific mySpin 6 Portable Centrifuge Kit 
CENT DNA other
Application note: Influenza virus purification using the Thermo Scientific Sorvall TCF-32 and CC 40 large-scale ultracentrifuge systems
CENT influenza other
Application note: Nanoparticle Separations in Thermo Scientific
Sorvall Micro-ultracentrifuges
CENT nano other
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.