Search Thermo Fisher Scientific
General Purpose Pro Centrifuges are designed to meet the needs of today’s rapid-fire discoveries, with updates to help you get your work done more quickly, consistently, and with powerful reliability.
CTS Series Equipment Packages consist of the equipment, a documentation package and compliance services. These packages support your GMP needs for cell and gene therapy manufacturing. They help you get up and running faster, stay compliant, support regulatory audits, and stay on schedule.
The General Purpose Pro Centrifuges are a refrigerated 4L floor and bench models equipped with an intuitive touchscreen interface, Auto-Lock Rotor Exchange Technology and Biocontainment Lids.
A comprehensive product documentation package includes regulatory certificates, recommended protocols and product-specific factory documentation and specification.
Gain confidence with our compliance services that can help reduce your risk of non-compliant instruments. Service options include Installation Qualification (IQ), Operation Qualification (OQ) and cycle testing.
Models | Sorvall X4R Pro | Sorvall X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Multifuge X4R Pro | Multifuge X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Sorvall X4R Pro | Sorvall X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Models | Multifuge X4R Pro | Multifuge X4F R Pro | ||||||||
![]() | ![]() | |||||||||
|
| |||||||||
Model | • Benchtop | • Floor standing | ||||||||
Capacity | • 4L capacity | • 4L capacity | ||||||||
Rotor | → TX-1000 rotor | → TX-1000 rotor | ||||||||
Buckets | → 4 x 1000mL | → 4 x 1000mL | ||||||||
Lids | → Biocontainment Certified Lids | → Biocontainment Certified Lids | ||||||||
Adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters | → 50 ml conical adapters → 15 ml conical adapters → 500 mlconical adapters |
Compliance Services* (separate order number required) – Reduce equipment noncompliance by documenting functionality to support your regulatory requirements with qualification (IQ/OQ) and performance testing services.
Qualification service Cat. No.
On-site IQ/OQ service IOQPCKE89003492
On-site IQ/OQ + cycle test IOPQPCKE89003492
*Compliance services are only available in select countries including U.S., Germany, France, U.K., Ireland, Spain, Portugal, Italy, Austria, Switzerland, Norway, Sweden, Denmark, Netherlands. Please state your interest to the “Request a quote” form.
Discover the Sorvall X Pro touchscreen features including:
Helps save time for faster results:
• Fiberlite rotor/lid combinations offer protection to the user, centrifuge and samples during centrifugation*
• Carbon fiber construction supported by a 15 year Fiberlite Rotor warranty
• Fiberlite Rotors reach higher speeds than metal counterparts, improving separations and results
• Ergonomics driven by lightweight construction, integrated handle design and Auto-Lock Rotor exchange
*Biocontainment certified by National Institute for Health Protection, Porton Down, UK
One-handed sample protection