Measuring protein concentration
Accurately quantifying total protein concentration is a key step in most experiments and workflows involving isolation, separation, and analysis of proteins. We offer a variety of assay reagents, kits, and standards for protein quantitation by fluorescent or colorimetric detection with fluorometers, spectrophotometers, and plate readers.
Additionally, our products are offered with Dilution-Free protein standards for added ease-of-use with reduced set up time when generating your standard curve. Dilution-Free protein standards are a set of prediluted standards, packaged in a multichannel pipette compatible format which allows you to easily transfer a full dilution series to your microplate, at once.
Which protein assay is right for you?
 packaging, reagent, and dilution-free tubes for Bradford assay
NEW Pierce Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standard
packaging, reagents, and dilution-free tubes for BCA protein assay
NEW Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards
packaging, reagents, and dilution-free tubes for BCA protein assay
NEW Pierce Dilution-Free Rapid Gold BCA Protein Assay Kit
packaging and reagents for qubit assay
Qubit Protein Broad Range Assay
Choose when
  • Speed and ease-of-use are important
  • Accuracy and reproducibility are important
  • Accuracy and reproducibility are important
  • Speed and ease-of-use are important
  • Compatibility and convenience are important
  • High-throughput is not important
Best-in-Class Accuracy (low protein-protein variation) 
Broad Dynamic Range (>2 mg/mL wide)  
Fast (≤ 15 min.) 
Dilution-Free Standard Curve Preparation
Dilution-Free Sample Preparation
High-Throughput (compatible with microplate format)
Compatible with Detergents 
Compatible with Reducing Agents
Automatic Concentration Calculations with Qubit 4
Automatic Concentration Calculations with SkanIt Software* 
*SkanIt software available for Multiskan FC, Multiskan SkyHigh, Varioskan ALF, and Varioskan LUX microplate readers. Learn more
We now offer Dilution-Free products, comprising both protein assays and protein standards that make protein quantification easy, fast, and accurate. Pierce Dilution-Free products allow you to minimize tedious dilutions and pipetting errors in your protein assays, delivering accurate results while reducing assay setup time by up to 80%. Learn more about our Dilution-Free assays.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
      Overview of assays
      packaging, reagents, and dilution-free tubes for BCA protein assaypackaging, reagent, and dilution-free tubes for Bradford assaypackaging and reagents for qubit assay
      BCA Assays
      Copper-based protein assays, including the bicinchoninic acid (BCA) and Lowry methods.
      • Compatible with most surfactants—even if present in the sample at concentrations up to 5%
      • Linear response curve (R2 > 0.95)
      • Less protein–protein variation than the Coomassie dye–based assays
      • Dilution-Free BSA protein standards for faster assay time and effortless standard curve generation
      View all BCA protein assays
      Bradford Assays
      Coomassie dye-binding assays for fast and simple protein quantification.
      • Typically faster and easier to perform compared to most protein assays
      • Performed at room temperature
      • Compatible with most salts, solvents, buffers, thiols, reducing substances, and metal-chelating agents
      • Dilution-Free BSA protein standards for faster assay time and effortless standard curve generation
      View all Bradford assays and reagents
      Fluorescent Protein Assays
      Fluorescence-based protein quantification detection methods provide superior sensitivity.
      • Excellent sensitivity, requiring less protein sample for quantitation
      • Timing is not a critical factor, so the assays can be adapted for automated handling in high-throughput applications
      • Good protein-to-protein variation
      View all fluorescent protein assays
      Related products and downstream applications
      Resources
      Pierce Protein methods – an encyclopedia of articles about protein analysis
      Protein assay technical guide – tools and reagents for improved quantitation of total or specific proteins.
      Protein assay compatibility table – summarizes compatible substances for several popular protein assays.
      Protein Analysis Learning Center – resources for different protein analysis techniques
      Protein Biology Application Notes – journal-style articles based on research applications and proof-of-concept experiments
      Liberate yourself with Pierce Dilution-Free protein assays
      For Research Use Only. Not for use in diagnostic procedures.