Fulfills a wide range of application requirements
The iBright Imaging Systems offer up to five imaging modes to support your multiple application requirements. Efficiently and easily capture data from protein gels, nucleic acid gels, chemiluminescent western blots, fluorescent western blots, and more.
Core imaging applications
Fluorescent western blots
4-plex multiplex fluorescent western blot imaged on iBright Imaging System
Chemiluminescent western blots
Multiplex chemiluminescent western blot imaged on iBright Imaging System
Combined fluorescent and chemiluminescent western blots
Multiplex chemiluminescent and fluorescent western blot imaged on iBright Imaging system
Colorimetric western blots
Colorimetric western blot imaged on iBright Imaging System
Fluorescent stained nucleic acid gels
Fluorescently stained nucleic acid gel imaged on iBright Imaging System
Fluorescent stained protein gels
Fluorescently stained protein gel imaged on iBright Imaging System
Colorimetric stained protein gels
Coomassie stained gel imaged on iBright 1500 Imaging System
Colorimetric membrane stains
Colorimetric stained western blot membrane imaged on iBright Imaging System
Specialty plate-based imaging applications
Fluorescent colonies (e.g. GFP expression)
GFP colonies, 470x370
Visible colonies (e.g. crystal violet stained)
Visible Crystal Violet Colonies, 470x370
Qualitative visible imaging applications*
TLC-plates
TLC plate, 470x370
Leaf sections
Leaf False Colored Green, 470x370
Images pictured for fluorescent western blots, stained nucleic acid gels, colorimetric stained protein gels, GFP expressing colonies, visible stained colonies, and leaf sections are shown in pseudocolor (false color applied). Data is captured in grayscale.* Note that qualitative visible imaging applications provide a qualitative visualization of the object or confirmation of the presence of signal and are not recommended for quantitation.
Optimized E-Gel precast agarose gel imaging and analysis
Leveraging the iBright Tray Adapters for E-Gel Agarose Gels helps streamline the image capture and analysis of our 11, 22, 48, and 96-well E-Gel configurations through appropriately centering the E-gels upon the iBright transilluminator and by blocking the E-gel label or barcode from contributing non-specific fluorescence.
Invitrogen iBright Tray Adapters for E-Gels.
The Invitrogen iBright Tray Adapter for E-Gels, 11/22-well (panel A) and the Invitrogen iBright Tray Adapter for E-Gels, 48/96-well (panel B).
Visual depiction of iBright Tray Adapters for E-Gels
The iBright Tray Adapter for 48/96-well E-Gel precast agarose gels on the iBright turntable.
The top component of the tray adapter (1) prevents fluorescence emitted by the E-Gel cassette (2) from reaching the camera detector. The bottom component of the tray adapter (3) centers the E-Gel cassette over the transilluminator (4) which is below the turntable (5). Light emitted by the green LED transilluminator shines through the open window in the bottom component.
Visual depiction of how the iBright tray adapters for E-gels fit on top of the iBright transilluminator.
Imaging with the iBright Tray Adapter for 48/96-well E-Gel precast agarose gels.
Gel imaged without tray adapter (left) and with the tray adapter (right).
The barcode and labeling at the top of the E-gel cassette in Panel A is covered by the iBright Tray Adapter for E-gels in Panel B which allows for the Smart Exposure algorithm to function properly.
Furthermore, our deconvolution workflow greatly simplifies the interpretation of high-throughput 96-well E-gel results because deconvolution makes it easier to review the data from lane-to-lane to better understand the results.
Touchscreen interface
The 12.1-inch LCD touchscreen interface has a simple, logical layout of functions and features, making our systems easy to use with minimal training. Workflows are similar between imaging modes, delivering a smooth imaging experience regardless of sample type.
Interface features
  1. Sign in: create a new user account of sign in to an existing account
  2. Gallery: access previously captured images
  3. Mode selection: dropdown menu to select imaging modes (chemiluminescent blot, fluorescent blot, nucleic acid gel, protein gel, and universal mode)
  4. Camera lock: maintain camera settings across multiple samples, locks exposure time, zoom, and transillumination settings while disabling mechanical auto-rotation
  5. Help: on-board help and information
  6. Settings: access general instrument settings, configurations, and service tools
  7. Drawer: open or close the sample drawer
  8. Exposure dial: select or turn the dial to set manual exposure time or fine-tune exposure time
  9. Capture: acquire images using exposure time set by user or instrument
  10. Edit channels: open multiple channel options to review and edit each channel
  11. More options: open image adjustment options
  12. Live view: go back to live-streaming sample view
  13. Analyze: initiate analysis workflow using current images
  14. Export: edit image information and export current images to destinations of choice
  15. Trash: delete current images
  16. Split screen: select one-window or two-window image view
  17. Color option: select color or grayscale image view
  18. Image view window: display selected image in the window for user view and interaction
Interface features
Powerful camera technology
Camera lens
The 9.1-megapixel cooled CCD camera captures crisp, clear, publication-quality images. High resolution enables more binning (pixel combining) options which provides flexibility for adjusting resolution and sensitivity based on need.
Smart Exposure technology rapidly determines optimal exposure time, minimizing the potential for over or underexposed images and the need to repeat exposures to get the desired signal.
Comparison of Smart Exposure technology to manually set exposure times.
Minimal pixel saturation is observed in the image captured with exposure time determined by Smart Exposure technology, while the range of data captured is maximized. The set of images on the right is the same as the image on the left, but with the saturated pixels feature of iBright Imaging Systems turned on (saturated pixels are displayed in red). The same blot was imaged using the exposure time determined by Smart Exposure technology, or four manually set exposure times.
Western blot data comparison between blot images captured with Smart Exposure technology compared to manually set exposure times.
For chemiluminescent western blot samples with widely varying expression levels, Smart Range HDR (high dynamic range) technology can help maximize the linear dynamic range. This feature leverages two different exposures of the same sample, a short exposure for capturing medium-to-high abundant proteins and a long exposure for capturing low abundant proteins. After capture, the two different images will be combined into a single 16-bit HDR image that contains both the medium-to-high and low abundant signal intensities to effectively extend the linear dynamic range beyond what is achievable with a single short or single long exposure time.
SmartRange HDR improved the detection limit of p23 4-fold compared to the Smart Exposure feature.
HeLa lysate was serially diluted 1:2 in sample buffer (20 µg, 10 µg, …10 ng), prepared for SDS-PAGE and electrophoresed on a Novex WedgeWell 4 to 20% Tris-Glycine gel. The protein was transferred to nitrocellulose membrane and probed for p23. The resulting Western blot was imaged on the iBright Imager using Smart Exposure and SmartRange HDR. 
Western blot data comparison and resulting graphical representation of the the linear dynamic range of blots captured with Smart Exposure technology compared to Smart Range HDR based image capture.
Automation streamlines operation
Mechanical rotation vs digital rotation and illustration of automatic mechanical rotation in the iBright Imager.
The iBright 1500 Series Imaging Systems automatically determine the sample position and can rotate samples left or right up to 10° on a mechanically rotating sample stage. This automation eliminates the need for repeated opening of the sample drawer to reposition your sample to achieve proper alignment.  In addition, mechanical rotation eliminates the need to digitally rotate the sample, which preserves the integrity of the data.
Digital rotation vs. mechanical rotation. (A) Pixels rotate with digital rotation so bands appear jagged. With mechanical rotation, the sample itself rotates, so bands remain smooth in appearance as the pixels remain aligned. (B) Graphic depicting iBright Imaging System sample stage before and after rotation.
In addition, iBright Imaging Systems automatically adjust the focus for each level of zoom, to maximally utilize the 22.5 cm x 18.0 cm field of view. If imaging a single blot, the camera will automatically zoom up to 2X zoom (1-2X zoom is mechanical zoom with iBright 1500 Series Imaging Systems, 1-2X zoom is digital zoom with the iBright CL750 Imaging system).  Mechanical zoom maximizes sensitivity by moving the camera closer to the sample stage and thus reduces focal length. The iBright 1500 Series Imaging Systems also provide additional 1-4X digital zoom for a combined zoom level of 1-8X.
Illustration of the 8x zoom function in iBright Imaging Systems using a multiplex fluorescent western blot.
Zoom function. (A) Unzoomed image of a fluorescent western blot. (B) Blot at 2X zoom. (C) Blot at 4X zoom. (D) Blot at 8X zoom. (blot not repositioned during successive zooms)
Large field of view in a small footprint
Illustration of iBright Imaging system field of view using 4 mini multiplex fluorescent western blots.
iBright tray open
The large field of view (22.5 x 18.0 cm) enables capture of up to 4 mini blots or gels.
Accelerate your work with fluorescent multiplexed western blots
Expand the possibilities—The iBright FL1500 model features five fluorescence channels, permitting up to 4-color fluorescent western blot multiplexing and expanding your possibilities for studying multiple proteins in a single blot. Obtain meaningful and representative comparisons to enhance your experiments. Smart Exposure technology further improves acquisition of multiplex fluorescent western blot data by ensuring that signal-to-noise ratios are optimized for each fluorescent channel separately.
Illustration of the fluorescent western blot imaging features in iBright FL1500
Green-LED transilluminator
iBright Imaging System green-LED transilluminator for nucleic acid gel imaging
The iBright Imaging Systems utilize a transilluminator based on green LEDs, which effectively excite popular DNA dyes such ethidium bromide and SYBR Green dye and offers several additional benefits.
No harmful UV rays: While UV light effectively excites many fluorescent dyes and stains, UV light is a health hazard. Further, prolonged exposure to UV light can damage DNA samples, and compromise the integrity of samples to be used for downstream applications, such as subcloning.
No mercury waste: UV transilluminator bulbs may contain mercury, a hazardous substance, and therefore require special care for handling and disposal.
Longer lifetime: LED bulbs have a substantially longer real-time life than fluorescent UV bulbs, which can add up to considerable cost savings over the lifetime of the instrument.
Protein normalization workflow
To account for variability in samples due to unequal protein sample concentration, inconsistent sample loading onto the gel, and uneven transfer of protein from the gel to membrane, an extra step called normalization is often performed by comparing proteins of interest to internal housekeeping protein controls such as GAPDH, β-tubulin, β-actin, and more. Although housekeeping proteins have been the historically popular choice for normalization, newer strategies involving the comparison of the protein of interest to total lane protein have emerged as a potentially more accurate alternative.
To support the methods best suited for your experiment, iBright Imaging Systems and iBright Analysis Software provide multiple quantitation and normalization options to monitor or mathematically compensate for experimental or sample variability.
Quantitative western blot analysis using Invitrogen No-Stain Protein Labeling Reagent demonstrating linearity of protein normalization
Total protein normalization using the No-Stain Protein Labeling Reagent. Invitrogen™ Bolt™ 4-12% Bis-Tris Plus gels were loaded with HeLa lysate ranging from 10 to 50 µg and electrophoresed using MES running buffer. Proteins from the gels were transferred onto mini PVDF membranes using the Invitrogen™ iBlot™ 2 Gel Transfer Device with iBlot 2 Transfer Stacks (P0 protocol for 7 minutes). The PVDF membranes were washed twice for 2 min with 20 mL of ultrapure water on a rotating platform, whereupon they were labeled with 10 mL of a working solution of No-Stain Protein Labeling Reagent on a rotating platform for 10 minutes. The membranes were then washed 3 times for 2 minutes with 20 mL of ultrapure water on a rotating platform, followed by addition of Invitrogen™ primary antibodies against β-actin (Cat. No. AM4302), GAPDH (Cat. No. 398600), and α-tubulin (Cat. No. 138000), and Invitrogen™ Goat Anti–Mouse IgG Alexa Fluor™ Plus 680 secondary antibody (Cat. No. A21058). The blot was imaged using the Invitrogen™ iBright™ FL1500 Imaging System. The iBright software was used to quantitate the total protein signal in the lanes. The R² value of the plotted data for the entire range of total protein signal was determined to be 0.9990, whereas the R² values for β-actin, GAPDH, and α-tubulin were 0.8851, 0.9438, and 0.8332, respectively.