Knoevenagel Condensation Reaction
In 1894, German chemist Emil Knoevenagel reported that diethyl malonate and formaldehyde condensed in the presence of diethylamine to form a bis-adduct. He later discovered that the same type of bis-adduct was produced when formaldehyde and other aldehydes were condensed with ethyl benzoylacetate or acetylacetone in the presence of primary and secondary amines. In 1896, he conducted further experiments, reacting benzaldehyde with ethyl acetoacetate at 0°C using piperidine as the catalyst to form ethyl benzylidene acetoacetate as the single product. The reaction of aldehydes and ketones with active methylene compounds in the presence of a weak base to produce α,β-unsaturated dicarbonyl or related compounds is now known as the Knoevenagel condensation reaction.
One of the general features of this reaction is that aldehydes react much faster than ketones. Additionally, the active methylene groups require two electron withdrawing groups, with typical examples including malonic esters, acetoacetic esters, malonodinitrile, or acetylacetone. Both the solvent and the nature of the catalyst employed are important. As the by-product of the reaction is water, removing the generated water by azeotropic distillation or by the addition of molecular sieves helps to shift the equilibrium to favor the formation of the product.
Mechanism of the Knoevenagel condensation reaction
Review available Thermo Scientific products for the Knoevenagel condensation reaction:
Catalog # Name Size Price (USD) Qty
A15468.36 Diethyl malonate, 99% Each
35.65
Special offer
Online exclusive
Ends: 30-Jun-2025
41.50
Save 5.85 (14%)
A15468.0E Diethyl malonate, 99% Each
92.65
Special offer
Online exclusive
Ends: 26-Sep-2025
109.00
Save 16.35 (15%)
A15468.0C Diethyl malonate, 99% Each
321.65
Special offer
Online exclusive
Ends: 26-Sep-2025
378.00
Save 56.35 (15%)
A16163.AP Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol Each
27.65
Special offer
Online exclusive
Ends: 30-Jun-2025
32.10
Save 4.45 (14%)
A16163.0F Formaldehyde, 37% w/w aq. soln., stab. with 7-8% methanol Each
52.65
Online Exclusive
58.00
Save 5.35 (9%)
L05030.18 Ethyl benzoylacetate, 90+% Each
35.65
Online Exclusive
39.90
Save 4.25 (11%)
L05030.30 Ethyl benzoylacetate, 90+% Each
75.65
Special offer
Online exclusive
Ends: 26-Sep-2025
88.40
Save 12.75 (14%)
L05030.0B Ethyl benzoylacetate, 90+% Each
238.65
Special offer
Online exclusive
Ends: 26-Sep-2025
281.00
Save 42.35 (15%)
A12544.30 Ethyl acetoacetate, 99+% Each
24.65
Special offer
Online exclusive
Ends: 26-Sep-2025
28.70
Save 4.05 (14%)
A12544.36 Ethyl acetoacetate, 99+% Each
29.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.10
Save 5.45 (16%)
A12544.0E Ethyl acetoacetate, 99+% Each
84.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.90
Save 14.25 (14%)
A12544.0C Ethyl acetoacetate, 99+% Each
289.65
Special offer
Online exclusive
Ends: 30-Jun-2025
340.00
Save 50.35 (15%)
A12442.AE Piperidine, 99% Each
35.60
A12442.AP Piperidine, 99% Each
77.50
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!  
Vote now