Separate polar compounds
Separate polar compounds
Is your sample insoluble in mixtures of water and organic solvent? We offer a range of bonded and non-bonded normal phase columns that are ideal for the retention and separation of lipophilic compounds.
Contact us
Normal phase HPLC column categories
Need to separate isomers using normal phase chromatography? These non-bonded silica columns are a powerful and efficient tool for separating non-polar and moderately polar isomeric compounds.
Using aqueous normal phase chromatography? In normal phase applications, amino phase columns offer alternative selectivity to silica. They are particularly useful for the retention and separation of carbohydrates.
Want to run normal phase chromatography with gradient elution? Cyano columns have alternative selectivity and equilibrate more quickly, offering greater suitability for normal phase gradient separations compared to unbonded silica columns.
Chromatography Consumables Resources and Information
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Discover more resources
2022/2023 HPLC/UHPLC Columns Catalog
Access the latest interactive version of our Chromatography Consumables & Columns Catalog.  Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
Get the HPLC/UHPLC columns catalog
Discover live and on-demand webinars that enable you to improve productivity, maintain compliance, and learn about the constantly evolving scientific landscape.
Watch now
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD SchemaApp code