Organism TypeMinkDerived from fetal lung; susceptible to vaccinia, herpes simplex, reovirus 3, vesicular stomatitis. ATCC CCL-64
TissuesLung
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,Stable Cell Transfection,Transfection: Gene Expression,Transient Transfection
Products
Catalog # Name Size Price (USD) Qty
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
842.65
Online Exclusive
924.00
Save 81.35 (9%)
11875093 RPMI 1640 Medium, 500 mL Each
37.37
18292011 Lipofectin™ Transfection Reagent, 1 mL Each
710.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.