Columns developed with your applications in mind
Our broad portfolio of ion chromatography (IC) columns offers an array of features from small particle sizes to high capacity capabilities, to the ability to accommodate dual eluent generators. The majority of our IC columns are available for convenient online purchase.
Small particle sizes available
Several of our ion exchange columns are available packed with 4µm resin particles. This smaller particle size provides better resolution, faster run time, and more reliable results. The length of the column can be optimized for the desired results. Shorter columns can be used at higher flow rates to increase productivity without sacrificing performance, while longer columns can be used for higher resolution separations of complex sample matrices. These options allow you to optimize resolution and throughput for your application.
4 µm particle-size columns
Benefits of using 4 µm particle-size columns.
Image of Dionex IonPac 4 micron particle size columns
High capacity
Our highly porous column resins offer more surface area per particle, providing more opportunity for analytes to bind to the resin surface. This high capacity enables the columns to handle high analyte concentrations, provide good resolution of weakly retained ions, enable analysis of high-ionic-strength samples, and load a larger sample volume without overloading and peak broadening. 
Column properties that make an impact on ion chromatography
Image of Dionex ion chromatography columns on a swirling blue background
Column selection
Our selection of Thermo Scientific Dionex ion chromatography columns ensures that we have the ideal columns for almost any type of analysis. We offer columns that can be used with hydroxide, carbonate and methanesulfonic acid (MSA) eluents. In addition to our Thermo Scientific Dionex IonPac IC columns, we offer unique column chemistries for high-resolution, high-efficiency analysis of biological molecules, such as amino acids and carbohydrates.
Dionex BioIC column selection guide
Choose the right IC column for your amino acid or carbohydrate analysis.
Image of selection guide for Thermo Scientific Dionex Bio-IC columns featuring three columns on a colorful background
Dual Eluent Generation Cartridge (EGC) compatibility
Our IC column portfolio includes columns for our Thermo Scientific Dionex ICS-6000 HPIC System that support running the system in Dual EGC mode. This mode uses electrolytic eluent generation technology to create highly reproducible, accurate KOH/KMSA eluent gradients that resolve mono-, di- and oligosaccharides and sialic acids.
Online purchasing
We offer convenient online purchasing of our Dionex ion chromatography columns, allowing you 24/7 access to track your order status, check order history and view invoices online. Register to take advantage of account-specific pricing and web-only price promotions.
1x1 image pixel for data collection