Unique column chemistries to solve your application challenges
We offer a complete line of ion chromatography columns for hydroxide, carbonate, and methanesulfonic acid (MSA) eluents, including anion and cation exchange columns in addition to amino acid and carbohydrate analysis columns, concentrator columns, trap columns and more. Many of our IC columns are available in our innovative 4µm-particle size format for high-pressure separations with greater resolution for your ion exchange chromatography.
Highlights of our ion chromatography columns
Wide column variety
Our large offering of IC columns means you will find the right column for your analytical needs.
A collection of four different ion chromatography columns
Small particle size
Columns featuring 4µm particles offer better resolution, faster run times and more reliable results.
Image of an ion chromatogram, looking like a graph with distinct peaks
Carbohydrate analysis
Choose from Thermo Scientific Dionex CarboPac Columns packed with different phases to ensure the optimal separation for your carbohydrate sample.
Image of the molecular structure of sugar
Online purchasing
Conveniently order new ion chromatography columns directly from our website.
An image of an electronic tablet featuring different types of products that can be purchased online.
Fast analysis
Our shorter 150 mm columns allow you to perform analyses at higher flow rates to increase productivity without sacrificing performance.
Fast analysis
Select the right column
Use our selection tools to find the ideal ion chromatography column for the matrix you need to analyze and your analyte of interest.
An infographic depicting circles of different colors going into a filter and circles of just one color emerging
For Research Use Only. Not for use in diagnostic procedures.