SYBR Green dye-based master mixes for real-time PCR
When using dye-based chemistry for real-time PCR analysis, master mix choice matters. As the pioneers of SYBR Green dye quantification for qPCR, we have evolved our SYBR Green master mix formulations over many years into highly consistent reagents that deliver exceptional sensitivity and specificity on any qPCR instrument.
Compare SYBR Green vs TaqMan chemistries for qPCR analysis
Graphic of female scientist over blue background
SYBR Green master mix products
For optimal performance using SYBR green dye-based qPCR, we recommended using our latest formulation, PowerTrack SYBR Green Master Mix. For automated workflows that require 72-hour bench-top stability, we recommend PowerUP SYBR Green Master Mix.
PowerTrack SYBR Green Master Mix
PowerTrack SYBR Green Master Mix boosts reaction setup accuracy with an optional visual-tracking dye and simplifies primer optimization by accommodating a broad primer Tₘ range.
PowerTrack SYBR Green Master Mix Tubes and Box over blue background
PowerUP SYBR Green Master Mix
PowerUp SYBR Green Master Mix features 72-hour benchtop stability with a dual hot-start mechanism for excellent specificity.
PowerUp SYBR Green Master Mix Tube and Box over blue background
Compare PowerTrack and PowerUp SYBR Green master mixes
PowerTrack SYBR Green Master Mix
PowerUp SYBR Green Master Mix
Chemistry
2-step SYBR Green dye
2-step SYBR Green dye
Target input
cDNA
cDNA
Concentration
2X
2X
Polymerase Taq DNA Polymerase Dual-Lock Taq DNA Polymerase
Multiplexing and passive reference dye  With ROX: 1 target With ROX: 1 target
Cycling mode: run time
Fast: ~30 min
Standard: ~60 min
Fast: ~30 min
Standard: ~60 minutes
Sample tracking dye (optional) Yes No
Reaction benchtop stability 8 hours 72 hours
Carryover contamination control
With UNG
With UNG
Reverse transcriptase
(sold separately)
SuperScript IV Vilo Master Mix
High-Capacity RNA-to-cDNA™ Kit
Assay recommendations
Sequence detection primers
Sequence detection primers
Ordering information
Quick order below or visit product page
Visit product page
Other SYBR Green Master Mixes
Our legacy SYBR Green master mixes, including Power SYBR Green PCR Master MixSYBR Green PCR Master MixFast SYBR Green qPCR Master Mix, and SYBR Select Master Mix have long supported researchers with their qPCR analysis. However, for optimal results we recommend upgrading to our newest master mixes, which offer superior performance and compatibility with any qPCR instrument.
Master mix performance data
For reliably accurate qPCR data, our latest formulation, PowerTrack SYBR Green Master Mix, has been optimized to provide highly specific target amplficiation and excellent data reproducibilty over a wide dynamic rage.
Figure 1. Highly specific amplification
Real-time PCR was performed using universal human reference (UHR) cDNA and primers targeting PGK1 (phosphoglycerate kinase 1), ARL1 (ADP-ribosylation factor-like protein 1), and SNF8 (vacuolar-sorting protein). Reactions were run in quadruplicate using master mixes from different suppliers. PowerTrack SYBR Green Master Mix generated a single melt curve in 100% of the reactions, indicating specific target amplification.
Panel of qPCR melt curves showing target amplification using different SYBR Green Master Mixes
Figure 2. Excellent data reproducibility
PowerTrack SYBR Green Master Mix shows reproducibility over a wide dynamic range. Six assays (PGK1, ARL1,SNF8, DF, GAPDH, and Corf1 19) were run in quadruplicate with UHR cDNA generated from four different RT kits. Assays were run with a 6-fold dilution series and 400 nM primer concentration.
Dot plot showing data reproducibility for qPCR reactions run with PowerTrack SYBR Green Master Mix
Instrument compatibility
Applied Biosystems SYBR Green master mixes can be used with any real-time PCR instrument, from any manufacturer. Refer to your master mix user guide for recommended experimental run conditions.
Ordering information
Quickly order PowerTrack SYBR Green Master Mix below. To purchase other SYBR Green master mixes, visit the product page for ordering information.
Catalog # Name Size Price (USD) Qty
A46012 Each
119.65
Online Exclusive
127.00
Save 7.35 (6%)
A46109 Each
509.65
Online Exclusive
546.00
Save 36.35 (7%)
A46110 Each
935.65
Online Exclusive
1,002.00
Save 66.35 (7%)
A46111 Each
2,251.65
Online Exclusive
2,400.00
Save 148.35 (6%)
A46112 Each
4,310.65
Online Exclusive
4,615.00
Save 304.35 (7%)
A46113 Each
4,296.65
Online Exclusive
4,600.00
Save 303.35 (7%)