Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Our anti-cow secondary antibodies are affinity-purified polyclonal antibodies with well-characterized specificity for cow immunoglobulin classes and subclasses. They are available in host species including goat, rabbit, chicken, mouse and sheep, and in the following formats:
Our secondary antibodies are available as whole IgG. Each of these antibodies is affinity purified using its target antigen (the primary antibody class, subclass, and fragment) coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in high specificity and low background signal. Conjugated antibodies are affinity purified before labeling.
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
For Research Use Only. Not for use in diagnostic procedures.