Conceptual art of a box of FBS bottles over a cell
Enabling consistency with Gibco heat inactivated FBS and gamma irradiated FBS
Heat inactivated FBS and gamma irradiated FBS are both effective in helping ensure the safety and reliability of cell culture research, serving distinct purposes depending on the specific needs of your application.
Heat inactivated FBS is treated to deactivate complement proteins that can interfere with immune assays and other sensitive cell culture applications, making it excellent for studies involving immunological responses or where complement activity could skew results.
Gamma irradiated FBS is treated to inactivate a broader range of contaminants, including viruses and mycoplasma, supporting a greater assurance of viral inactivation.
View GI FBS  View HI FBS  View GIHI FBS  Try iMATCH Sera Lot Matching Tool
Gamma irradiated FBS: Enhanced sterility for reliable cell cultures
Gibco gamma irradiated fetal bovine serum (FBS) is designed to reduce viral risk by inactivating a broad range of contaminants, including viruses and mycoplasma. This treatment helps ensure safer and more consistent outcomes.
  • Enhanced viral inactivation: Reduces the risk of introducing contaminants into cell cultures.
  • Regulatory compliance: Gamma Irradiation is recognized by regulatory agencies as the best method of viral inactivation. Fully validated for viral inactivation with doses ranging from 25 to 45 kGy.
Robust quality standards are maintained for Gibco gamma irradiated fetal bovine serum
Our gamma irradiation process helps to ensure consistency and traceability for a higher level of assurance when handling or using animal derived products.
Image of a bottle containing Gibco Gamma Irradiated FBS
When is gamma irradiation of serum necessary?
Gamma irradiation of serum enhances the safety and efficacy of animal-derived products by inactivating viruses and other pathogens detrimental to cell culture and biopharmaceutical applications. This process mitigates the risk of adventitious agent contamination, crucial in the production of vaccines and biopharmaceuticals. Furthermore, incorporating irradiated serum from the earliest stages of product development helps ensure consistency and minimizes viral risks.
Heat inactivated FBS: Reliable performance for immunological studies
Gibco heat inactivated fetal bovine serum (HI FBS) undergoes a controlled heat inactivation process to deactivate complement proteins, making it exceptional for immunological research.
  • Complement inactivation: Prevents interference in immune assays and cell cultures.
  • Validated process: Heated for 30 minutes at 56°C to inactivate complement proteins, helping to ensure reliability.
  • Trusted quality: Maintains robust quality standards, helping to ensure consistent experimental results.
Robust quality standards are maintained for Gibco heat inactivated fetal bovine serum
Results from biochemical, hormonal profile, bacterial, and fungal testing are virtually unchanged in Gibco heat inactivated FBS when compared to regular FBS products. You can expect the same robust quality standards to be met, including endotoxin and hemoglobin levels.
Image of a bottle containing Gibco Heat Inactivated FBS
When is heat inactivation of serum necessary?
Heat inactivation of fetal bovine serum can be used to inactivate complement proteins in the serum. Complement proteins are components of the innate immune system that contribute to inflammation and can opsonize pathogens. In addition to interacting with pathogens directly, complement proteins can also bind antibody-antigen complexes [1]. Interactions between complement proteins and antibodies may interfere with the binding of target molecules in immunoassays, making the inactivation of serum complement proteins a necessary step in certain immunological applications [2].
Importantly, heat inactivation of serum must be performed carefully to avoid the destruction of growth-promoting factors.
Catalog # Name Size Price (USD) Qty
A5209501 Fetal Bovine Serum, Value, heat inactivated, One Shot™ format, 50 mL Each
70.65
Online Exclusive
76.50
Save 5.85 (8%)
A5209502 Fetal Bovine Serum, Value, heat inactivated, One Shot™ format, 10 x 50 mL Each
663.65
Online Exclusive
722.00
Save 58.35 (8%)
10438034 Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions, 1000 mL Each
1,192.00
A5256801 Fetal Bovine Serum, Value, heat inactivated, 500 mL bottle Each
561.65
Online Exclusive
612.00
Save 50.35 (8%)
A5670801 Fetal Bovine Serum (FBS), Premium, heat-inactivated, 500 mL Each
801.65
Online Exclusive
900.00
Save 98.35 (11%)
A5670501 Fetal Bovine Serum (FBS), Premium, heat-inactivated, One Shot™ format, 50 mL Each
101.65
Online Exclusive
113.00
Save 11.35 (10%)
A5670502 Fetal Bovine Serum (FBS), Premium, heat-inactivated, One Shot™ format, 10 x 50 mL Each
943.65
Online Exclusive
1,060.00
Save 116.35 (11%)
A5669801 Fetal Bovine Serum, Premium Plus, heat inactivated, 500 mL bottle Each
945.65
Online Exclusive
1,076.00
Save 130.35 (12%)
A5669501 Fetal Bovine Serum, Premium Plus, heat inactivated, One Shot™ format, 50 mL Each
120.65
Online Exclusive
135.00
Save 14.35 (11%)
A5669502 Fetal Bovine Serum, Premium Plus, heat inactivated, One Shot™ format, 10 x 50 mL Each
1,125.65
Online Exclusive
1,268.00
Save 142.35 (11%)
Dual-treated option: gamma irradiated & heat inactivated FBS
For the highest confidence in safety and reliability, choose Gibco FBS that has undergone both heat inactivation and gamma irradiation. This dual treatment maximizes the safety of the serum, significantly reducing the risk of contamination and variability in high-stakes applications.
Contact an FBS representative
We have dedicated regional sales representatives to assist you in all things FBS. Connect directly with a FBS representative today!
Contact us
decorative image
References
Resources
Fetal Bovine Serum Basics
Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Gibco iMatch Sera Lot Matching Tool 
Provide your previous Gibco FBS lot number or answer a few questions to find your ideal serum match.
Related products
Support
Find your regional sales representative
Connect with a regional FBS sales representative.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of laboratory products.
For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Stylesheet for Classic Wide Template adjustments