Microplate Readers
Consistent results, exceptional user experience across many applications.
Thermo Scientific microplate readers offer flexibility, reliable performance, and ease of use for a variety of assays. Whether you need to measure absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF), or AlphaScreen assays, we can offer a microplate reader to meet the requirements of your specific workflow. 
Thermo Scientific plate readers have several features to help save time and maximize productivity, such as self-diagnostics, auto-calibration, and automation readiness with robot compatibility. Intuitive SkanIt Software makes microplate reading easy, providing excellent usability and flexibility, even for the most challenging microplate assays. SkanIt Software offers a visual workflow setup, an extensive library of ready-made protocols, and effortless data reduction and export. Explore our selection guide of dedicated modular and upgradable multimode readers, related products, assays, and accessories.
Video Player is loading.
Current Time 0:00
Duration 1:33
Loaded: 10.51%
Stream Type LIVE
Remaining Time 1:33
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    Conserve your precious samples with µDrop plates
    Ideal tools for quick and easy photometric, low-volume (2–10 μL) measurements of DNA, RNA, and protein concentration, or spectral scanning.
    Select from over 2000 ELISA kits
    Quickly and easily detect cytokines, proteins, and other markers with highly referenced ELISA kits.
    Accurate and precise nucleic acid quantification
    Choosing the right technology for your DNA and RNA samples can save significant time and money by preventing downstream experimental failures.
    Optimal washing performance
    Reliable and easy-to-use Wellwash microplate strip washers provide secure washing performance for ELISA applications.
    Fluorescence microplate assays
    From cell viability and proliferation assays to enzyme activity and protein quantification assays, these tools offer accurate and reliable measurements for kinetic and endpoint protocols across a wide range of biological applications.
    Quant-iT assay kits and reagents
    Quant-iT assay kits use fluorescent dyes which bind selectively to DNA, RNA, protein, or endotoxin. Our kits provide the ultimate convenience for quantitation in microplate assays.
    Microplate readers are everyday lab tools used extensively to perform microplate assays using single mode or multimode functions. Microplate readers can measure well plates with variable sample volumes, accommodating different microplate and microtiter formats based on the required sample volume. Thermo Fisher Scientific offers microplate readers for detection of absorbance, fluorescence intensity, luminescence, time-resolved fluorescence (TRF), and AlphaScreen microplate assays. Our multimode plate readers can read a variety of well plates to perform assays for both research and drug discovery screening applications.
    For Research Use Only. Not for use in diagnostic procedures.