High throughput screening with multiwell plates
Improved specificity for high-confidence screening results
Silencer Select siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications to yield siRNAs with unrivalled efficacy, potency, and specificity.
Improve RNAi screening results with highly efficient, specific siRNAs
The benefits of Silencer Select siRNAs are amplified in the context of high-throughput screening applications. Trust your discovery to the siRNA with our knockdown guarantee in the industry!
AttributesAdvantages
  • Advanced bioinformatic seed region analysis for sequence selection
  • LNA chemical modifications
  • Reduced off-target effects by up to 90% for fewer false positives
  • Less toxicity due to off-target effects
  • More consistent phenotypic data
  • Next-generation design algorithm
  • Up to 100-fold more potent silencing than other available siRNAs
  • Improved silencing for more robust knockdown results
  • Fewer false negatives for greater experimental confidence
  • Three unique individual siRNAs per gene
  • Multiple data points per gene for rapid progression from primary screening to hit confirmation studies
If you’re ready to start screening with a pre-defined collection or want to request a custom library, request a quote now!
Request a quote 
Pre-defined Silencer Select siRNA libraries
These libraries have been pre-assembled using PANTHER (www.pantherdb.org ) and Gene Ontology™ annotation information to create up-to-date target gene sets.
  • Each gene is targeted by three unique siRNAs. Validated siRNAs are included at no extra cost.
  • 0.25 nmol of each siRNA is provided dried-down in the wells of barcoded 96- or 384-well plates. This is sufficient for 100-500+ transfections, depending on siRNA working concentration and reaction volume.
  • Column 12 of each 96-well plate and columns 23 and 24 in each 384-well plate are left empty to allow for the addition of controls at the time of use.
  • Plates are conveniently arranged for pooling of siRNA, if desired.
  • The libraries are delivered with plate map files including full siRNA sequence information, as well as each targeted gene's symbol, full name, aliases, RefSeq accession, and Entrez Gene ID.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAisupport@thermofisher.com
Ordering information—Silencer Select siRNA libraries
Catalog # Name Size
A30085 Silencer™ Select Human Epigenetics siRNA Library Each
A30087 Silencer™ Select Human Transcription Factor siRNA Library Each
A30089 Silencer™ Select Human DNA Damage Response siRNA Library Each
A30091 Silencer™ Select Human Drug Targets siRNA Library (384 well), 1 set (384-well) Each
A30092 Silencer™ Select Human Drug Targets siRNA Library (96 well), 1 set (96-well) Each
A30095 Silencer™ Select Human Apoptosis siRNA Library Each
A30133 Silencer™ Select Human Drug Transporter siRNA Library Each
A30136 Silencer™ Select Human Cell Cycle Regulation siRNA Library Each
A30138 Silencer™ Select Human Membrane Trafficking siRNA Library Each
A30140 Silencer™ Select Human Ubiquitin siRNA Library Each
A30142 Silencer™ Select Human Cancer Genome siRNA Library Each
A30144 Silencer™ Select Human Cell Surface siRNA Library Each
A30146 Silencer™ Select Human Tumor Suppressor siRNA Library Each
A30148 Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library, 1 set (96-well) Each
A30149 Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library (384 well), 1 set (384-well) Each
4397914 Silencer™ Select Human Nuclear Hormone Receptor siRNA Library V4 Each
4397915 Silencer™ Select Human Ion Channel siRNA Library V4 Each
4397916 Silencer™ Select Human GPCR siRNA Library V4 Each
4397917 Silencer™ Select Human Protease siRNA Library V4 Each
4397918 Silencer™ Select Human Kinase siRNA Library V4 Each
4397919 Silencer™ Select Human Phosphatase siRNA Library V4 Each
4397926 Silencer™ Select Human Genome siRNA Library V4 Each
4397923 Silencer™ Select Human Genome siRNA Extension Set V4 Each
4397920 Silencer™ Select Human Druggable Genome siRNA Library V4, 1 set (96-well) Each
4397921 Silencer™ Select Human Druggable Genome siRNA Extension Set V4, 1 set (96-well) Each
4397927 Silencer™ Select Human Extended Druggable Genome siRNA Library V4 Each
Custom Silencer Select siRNA libraries
Silencer Select siRNAs are available as custom collections to suit your specific needs. These highly economical plated versions of our guaranteed siRNAs are perfect for pilot studies, primary screen follow-up, or a customized functional genomics screen.
  • Minimum order of 20 siRNAs
  • Up to 3 siRNAs per gene
  • Choose from 96-well, 384-well, or 384-well Echo-qualified plates
  • Validated siRNAs are available at no additional cost!
Product line for custom siRNA librariesQuantity per well*
Silencer Select human pre-designed inventoried siRNA 0.1, 0.25, 1, or 2 nmol
*NOTE: Not all quantities available in 384-well
Publications featuring Silencer Select siRNA libraries
Since 2010, Silencer Select siRNA libraries have been enabling functional genomics studies in labs around the world. These selected peer-reviewed references showcase the power of our high-quality siRNAs in high-throughput screening applications.
Pre-defined Silencer Select siRNA libraries
These libraries have been pre-assembled using PANTHER (www.pantherdb.org ) and Gene Ontology™ annotation information to create up-to-date target gene sets.
  • Each gene is targeted by three unique siRNAs. Validated siRNAs are included at no extra cost.
  • 0.25 nmol of each siRNA is provided dried-down in the wells of barcoded 96- or 384-well plates. This is sufficient for 100-500+ transfections, depending on siRNA working concentration and reaction volume.
  • Column 12 of each 96-well plate and columns 23 and 24 in each 384-well plate are left empty to allow for the addition of controls at the time of use.
  • Plates are conveniently arranged for pooling of siRNA, if desired.
  • The libraries are delivered with plate map files including full siRNA sequence information, as well as each targeted gene's symbol, full name, aliases, RefSeq accession, and Entrez Gene ID.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAisupport@thermofisher.com
Ordering information—Silencer Select siRNA libraries
Catalog # Name Size
A30085 Silencer™ Select Human Epigenetics siRNA Library Each
A30087 Silencer™ Select Human Transcription Factor siRNA Library Each
A30089 Silencer™ Select Human DNA Damage Response siRNA Library Each
A30091 Silencer™ Select Human Drug Targets siRNA Library (384 well), 1 set (384-well) Each
A30092 Silencer™ Select Human Drug Targets siRNA Library (96 well), 1 set (96-well) Each
A30095 Silencer™ Select Human Apoptosis siRNA Library Each
A30133 Silencer™ Select Human Drug Transporter siRNA Library Each
A30136 Silencer™ Select Human Cell Cycle Regulation siRNA Library Each
A30138 Silencer™ Select Human Membrane Trafficking siRNA Library Each
A30140 Silencer™ Select Human Ubiquitin siRNA Library Each
A30142 Silencer™ Select Human Cancer Genome siRNA Library Each
A30144 Silencer™ Select Human Cell Surface siRNA Library Each
A30146 Silencer™ Select Human Tumor Suppressor siRNA Library Each
A30148 Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library, 1 set (96-well) Each
A30149 Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library (384 well), 1 set (384-well) Each
4397914 Silencer™ Select Human Nuclear Hormone Receptor siRNA Library V4 Each
4397915 Silencer™ Select Human Ion Channel siRNA Library V4 Each
4397916 Silencer™ Select Human GPCR siRNA Library V4 Each
4397917 Silencer™ Select Human Protease siRNA Library V4 Each
4397918 Silencer™ Select Human Kinase siRNA Library V4 Each
4397919 Silencer™ Select Human Phosphatase siRNA Library V4 Each
4397926 Silencer™ Select Human Genome siRNA Library V4 Each
4397923 Silencer™ Select Human Genome siRNA Extension Set V4 Each
4397920 Silencer™ Select Human Druggable Genome siRNA Library V4, 1 set (96-well) Each
4397921 Silencer™ Select Human Druggable Genome siRNA Extension Set V4, 1 set (96-well) Each
4397927 Silencer™ Select Human Extended Druggable Genome siRNA Library V4 Each
Custom Silencer Select siRNA libraries
Silencer Select siRNAs are available as custom collections to suit your specific needs. These highly economical plated versions of our guaranteed siRNAs are perfect for pilot studies, primary screen follow-up, or a customized functional genomics screen.
  • Minimum order of 20 siRNAs
  • Up to 3 siRNAs per gene
  • Choose from 96-well, 384-well, or 384-well Echo-qualified plates
  • Validated siRNAs are available at no additional cost!
Product line for custom siRNA librariesQuantity per well*
Silencer Select human pre-designed inventoried siRNA 0.1, 0.25, 1, or 2 nmol
*NOTE: Not all quantities available in 384-well
Publications featuring Silencer Select siRNA libraries
Since 2010, Silencer Select siRNA libraries have been enabling functional genomics studies in labs around the world. These selected peer-reviewed references showcase the power of our high-quality siRNAs in high-throughput screening applications.
Questions?
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.