Short/Small Interfering RNAs
High quality siRNA reagents for potent gene silencing
RNA interference (RNAi) using small, interfering RNA (siRNA) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types.
  • Choose from guaranteed, pre-designed siRNAs targeting human, mouse, or rat
  • Validated siRNAs available against key human and mouse targets
  • Specificity-enhanced siRNA reduces the likelihood of off-target effects
Search pre-designed siRNA
Choose from our pre-designed siRNA product lines
 Silencer Select siRNAStealth RNAi siRNASilencer siRNA
 RECOMMENDED
Highest knockdown, fewest off-targets
Good knockdown, reduced off-targetsCost-effective, guaranteed siRNA
Potency (recommended working conc.) 5 nM20 nM50 nM
Silencing guarantee
Learn more
2 of 2 siRNA 2 of 3 siRNA2 of 3 siRNA
Target specificityHighestHighModerate
Chemical modifications, length
  • LNA-modified
  • 19-bp duplex
  • 2 nt overhangs
  • Modified
  • 25-bp duplex
  • no overhangs
  • Unmodified
  • 19-bp duplex
  • 2 nt overhangs
Gene types targetedProtein-coding genesProtein-coding genes
siRNA LibrariesNot available
Target speciesHuman, mouse, rat
(Other species: order custom designs)
Custom siRNA ordering toolCustom Silencer Select siRNACustom Stealth RNAi siRNA Custom Silencer siRNA
New to siRNAComplete the RNAi workflow with optimal siRNA delivery and tools for mRNA knockdown detection!
siRNA videos
Meet the inventor: Susan Magdaleno discusses Silencer Select siRNA technology
Dr. Gavin Robertson describes how Silencer Select siRNA technology is advancing melanoma research.
Shop for everyday lab essentials to power your transfection steps
Transfection lab essentials
We've made it easier for you to quickly find and buy products for every step in your transfection workflow.
Shop now
Questions?
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.