Search Thermo Fisher Scientific
Organism Type | Human | 293-H cells are GIBCO brand cells. These are clonal isolates derived from transformed human embryonal kidney (HEK) cells. The 293-H strain is a variant of the 293 cell line that demonstrates better adherence in monolayer culture and ease of use for plaque assays and other anchorage dependent applications. They have been adapted to suspension culture in serum-free media, 293 SFM II. |
Tissues | Kidney | |
Phenotype | Either | |
Primary | no | |
Application |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||