Organic extraction is an essential technique in molecular biology, commonly used for isolating nucleic acids, proteins, and cells from complex biological samples. This method relies on the differential solubility of biological molecules in organic solvents and aqueous phases, enabling the effective separation of these biomolecules. While spin columns and magnetic bead-based extraction have grown in popularity, organic extraction remains a valuable approach in many research applications due to its efficiency and cost-effectiveness.
Learn more: DNA protocols
Explore: Bulk reagents
Core principles of organic extraction
The core principle behind organic extraction is the partitioning of biomolecules into different phases based on their chemical properties. This typically includes an aqueous phase and an organic phase. Organic solvents, such as phenol and chloroform, are added to biological samples, creating a biphasic system. Proteins and lipids tend to partition into the organic phase, while nucleic acids are hydrophilic and remain in the aqueous phase. This process takes advantage of differences in solubility and hydrophobicity, enabling the efficient isolation of desired analytes.
Buffers and pH adjustments are critical in this technique. For example, when extracting DNA, the pH is adjusted to be neutral to slightly basic at around 7–8. This helps ensure the DNA remains in the aqueous phase, while for RNA, acidic conditions are used to keep RNA in the aqueous phase and DNA in the organic layer. After phase separation by centrifugation, the aqueous phase containing nucleic acids can be easily collected, while proteins and other contaminants remain in the organic phase or the interphase.
Organic purification methods based on biomolecule
The choice of purification method depends on the specific properties of the target biomolecule, including its size, charge, solubility, and affinity for certain ligands. Effective biomolecule purification is essential for downstream applications, including structural analysis, functional studies, and therapeutic development. Three main biomolecules studied are nucleic acids, proteins, and cells.
Organic method of DNA and RNA extraction
Nucleic acid purification icon
The most common method for nucleic acid extraction is the phenol-chloroform extraction technique. In this method, phenol, often mixed with chloroform and isoamyl alcohol, is added to the biological sample. Upon centrifugation, the nucleic acids partition into the aqueous phase, while proteins and lipids move into the organic phase or precipitate at the interphase.
The pH of the phenol is crucial in determining whether DNA or RNA is selectively extracted. For DNA extraction, neutral or slightly alkaline conditions are used, while for RNA extraction, an acidic pH favors RNA recovery in the aqueous phase. Once the aqueous phase is collected, nucleic acids are typically precipitated with ethanol or isopropanol and washed to remove any residual solvents.
This method is commonly used in genomic DNA extractionRNA purification, and plasmid isolation, especially when high purity and yield are necessary for downstream applications such as cloning or sequencing.
Organic extraction of proteins
Protein icon
Proteins can be extracted and purified using organic solvents such as phenol-acetone or trichloroacetic acid (TCA). In protein extraction, organic solvents denature proteins, precipitating them out of the aqueous solution. For example, TCA precipitation is often followed by washing with organic solvents such as acetone to remove salts and other contaminants.
Protein extraction via organic solvents is particularly useful in proteomic studies, where high-quality proteins are needed for mass spectrometry analysis or enzyme assays. This method also allows for the removal of nucleic acids and other unwanted biomolecules that might interfere with protein analysis.
Organic extraction of cells (cell fractionation)
Cell icon
In cell extraction and fractionation, organic solvents combined with detergents are often used to lyse cells and release cellular components. Detergents like SDS or Triton X-100 disrupt the cell membrane, while organic solvents help solubilize the membrane lipids. By selectively extracting specific cell components, such as membranes or organelles, researchers can study individual parts of the cell in greater detail.
This method is essential for isolating cellular structures like mitochondria, membranes, and other organelles for biochemical and structural studies.
Featured products
TRIzol RNA isolation
TRIzol Reagent to deliver intact, high-quality RNA from their chosen biological samples.
Browse TRIzol reagents
Phenol-chloroform extraction
Phenol-chloroform is commonly used by molecular biologists to remove proteins from nucleic acids.
View phenol-chloroform protocol
RNase control
Prevent RNase and DNase contamination using a combination of clean surfaces and nuclease-free lab supplies.
Learn about RNase decontamination
Reagents commonly used in organic extraction
Organic extraction requires a range of reagents to facilitate the phase separation and isolation of biomolecules:
  • Phenol—an essential solvent that denatures proteins by disrupting hydrogen bonds, allowing nucleic acids to remain in the aqueous phase
  • Chloroform—enhances phase separation by promoting protein and lipid partitioning into the organic phase
  • Isoamyl alcohol—often used in conjunction with chloroform to reduce foam formation and stabilize the organic-aqueous interface
  • Ethanol/isopropanol—commonly used to precipitate nucleic acids or proteins after extraction, aiding in the recovery and purification of these molecules
  • Detergents (e.g., SDS)—solubilize cell membranes, facilitating the release of intracellular contents during cell lysis
View UltraPure Phenol:Chloroform:Isoamyl Alcohol
View Absolute Ethanol, 200 proof, Molecular Biology Grade
View UltraPure Sodium Dodecyl Sulfate (SDS)
Factors influencing organic extraction
Several factors must be optimized for successful organic extraction:
  • Sample type—the choice of reagents and method can vary depending on whether the sample is derived from cells, tissues, or bodily fluids. Different samples may require specific protocols to maximize yield and purity.
  • pH and buffer conditions—the pH of the extraction buffer determines which molecules are retained in the aqueous phase. For example, acidic phenol is used to extract RNA, while neutral phenol is used for DNA.
  • Organic solvent choice—the solvent used impacts the integrity of proteins and the recovery of nucleic acids. Chloroform, for example, can help efficiently separate lipids and proteins from nucleic acids.
  • Temperature and centrifugation speed—proper control of temperature and centrifugation speed helps ensure efficient phase separation and minimizes the risk of sample degradation
Pros and cons of organic extraction
There are several advantages and disadvantages in choosing organic extraction as a technique to isolate biomolecules from complex mixtures.
Advantages of organic extraction
Organic extraction is advantageous for yielding high-purity DNA from complex samples, while spin column extraction offers speed, convenience, and safety. Magnetic bead-based extraction stands out for its automation potential, scalability, and high purity, making it suitable for high-throughput and diverse applications. The choice of method depends on the specific requirements of the experiment, including the type of sample, desired purity, and throughput needs. Additionally, organic extraction efficiently separates nucleic acids from proteins, lipids, and other cellular components and is relatively inexpensive, making this method affordable for large sample volumes.
Disadvantages of organic extraction
Organic solvents like phenol and chloroform are highly toxic and require careful handling and disposal. The organic extraction process is labor-intensive and involves multiple steps, making it more time-consuming compared to automated methods. If not carefully executed, cross-contamination between phases of organic extraction can reduce the purity of the isolated biomolecules.
KingFisher purification instruments
Automated isolation, extraction, and purification of DNA, RNA, proteins, and cells. Applied Biosystems MagMAX optimized kits and reagents enable easy-to-use protocols that remove manual steps and help save time.
Learn more
Applications of organic extraction in molecular biology
  • Molecular cloning—high-quality DNA is critical for molecular cloning techniques, where phenol-chloroform extraction remains widely used
  • Proteomics—extracting proteins with minimal nucleic acid contamination is essential for proteomic analysis, enzyme assays, and structural studies
  • Transcriptomics and RNA-seq—RNA extraction using organic solvents is crucial for isolating high-quality RNA for gene expression profiling and transcriptomic studies
  • Cell biology—organic solvents play a key role in cell fractionation, allowing researchers to isolate organelles and membranes for functional studies
Troubleshooting common issues in organic extraction
  • Poor phase separation—inadequate centrifugation or incorrect solvent ratios can prevent proper phase separation. Optimizing centrifugation conditions and solvent ratios can help resolve this issue.
  • Low yield—to maximize nucleic acid or protein yield, check if the correct buffer conditions and solvent volumes are being used
  • Contamination—if DNA or RNA is contaminated with proteins, additional washing steps can help remove contaminants
  • Degraded samples—proper storage, handling, and the use of fresh reagents are crucial to prevent sample degradation
Future directions and innovations in organic extraction
The field of organic extraction is evolving, with new trends focused on improving safety, efficiency, and automation:
  • Automation—high-throughput systems are integrating organic extraction protocols, making it easier to process large numbers of samples efficiently
  • Greener reagents—there is increasing interest in developing less toxic and more sustainable organic solvents to replace phenol and chloroform
  • Improved protocols—novel buffer systems and organic solvent alternatives are being explored to enhance extraction from challenging samples such as low-yield tissues or difficult-to-lyse cells
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.