Molecular Biology Bulk Reagents and Lab Supplies for Sample Preparation
Sample preparation for molecular biology research requires essential reagents and lab supplies that may include buffers, enzymes, primers, RNase-free tubes, tips and plates, centrifuges, thermocyclers, and protective lab equipment. Researchers and lab managers can stock up on reagents and lab supplies for sample preparation with Thermo Fisher. While performing experiments or running tests at larger scales and sample sizes, ordering bulk reagents helps reduce the cost of laboratory operations. 
  • DNA polymerases
  • RNAse control
  • TRIzol RNA isolation
  • Nuclease-free water
  • RNase-free tubes and pipette tips
  • Multi-well plates
  • Centrifuge spin columns
  • Latex, nitrile and vinyl gloves
  • Bulk and OEM magnetic beads
  • Scalable Specialty chemical services 
Bulk reagents for sample preparation
TRIzol RNA isolation
TRIzol Reagent is a patented formulation that supports the extraction of total RNA from tissues or cells, including lipid-rich and difficult sample types. The product has been used in countless labs and referenced in thousands of journal publications as labs use TRIzol Reagent to deliver intact, high-quality RNA from their chosen biological samples.
Phenol chloroform extraction
Phenol-chloroform is commonly used by molecular biologists to remove proteins from nucleic acids. The phenol is premixed with chloroform:isoamyl at a ratio of 25:24:1. 
Cells-to-CT
Cells-to-CT Bulk RT Reagents are large quantity reagents used for reverse transcription (RT) in the TaqMan Fast Advanced Cells-to-CT Kit and the SYBR Green Fast Advanced Cells-to-CT kit. 
RNAse controls
Prevent RNAse and DNase contamination using a combination of clean surfaces and nuclease free lab supplies. Improve the purity of your final sample by improving the control. Once you master the basics of RNAse control, you are ready to browse our selection of essentials.
RNAse control
DNA polymerases
DNA polymerases are enzymes that are essential for DNA replication, repair, and amplification. Bulk DNA polymerases are available at a lower cost, allowing researchers to purchase larger quantities and reduce the cost of laboratory operations while helping ensure consistency and reliability of results.
Nuclease-free water
Use nuclease-free water that’s used to prepare reagents or to resuspend precipitated RNA. We offer four primary formats of nuclease-free water: 
  • DEPC-treated
  • Non DEPC-treated
  • RT-PCR grade water
  • DNase-/RNase-free water
Catalog # Name Size Price (USD) Qty
16096040 Each
634.65
Online Exclusive
662.00
Save 27.35 (4%)
15596018 Each
493.00
10296028 Each
594.00
A33251 Each -
15593049 Each
485.65
Online Exclusive
512.00
Save 26.35 (5%)
17909 Each
318.00
A39110 Each
10,690.00
A35378 Each
9,160.00
A25602 Each
7,193.65
Online Exclusive
7,870.00
Save 676.35 (9%)
AM2696 Each
604.00
AM2684 Each
550.65
Online Exclusive
608.00
Save 57.35 (9%)
EP0042 Each
378.65
Online Exclusive
445.00
Save 66.35 (15%)
EP0094 Each
1,096.65
Online Exclusive
1,316.00
Save 219.35 (17%)
EP0162 Each
353.65
Online Exclusive
435.00
Save 81.35 (19%)
4387937 Each
258.65
Online Exclusive
274.00
Save 15.35 (6%)
4387936 Each
281.00
10977023 Each
268.00
Lab supplies for sample preparation
Tubes and pipette tips
RNase-free tubes are indispensable for RNA-based applications. These tubes are treated to inactivate RNase, an enzyme that degrades RNA, thus helping ensure the integrity of RNA samples. They are commonly used in procedures such as reverse transcription, PCR, and RNA storage.
  • PCR tubes
  • Microcentrifuge tubes
  • Conical tubes
Multi-well plates
Multi-well plates from 6 well plates to 96 deep-well plates are used in so many life science workflows. Buying in bulk will reduce your cost and save you time on restocking your essential plastics. 
Multi-well plates
Centrifuge spin columns
Centrifuge spin columns are used for purifying, concentrating, and desalting biological samples. They have the power to separate DNA, RNA, and proteins from other cellular components by using centrifugal force to drive the sample through a filter or gel matrix, separating it based on size or other properties. Bulk centrifuge spin columns in larger quantities reduce the cost of laboratory operations and help ensure consistency and reliability of results.
 Centrifuge spin columns
Latex, nitrile and vinyl gloves
Gloves are an essential component of laboratory safety, protecting researchers from harmful chemicals, biological agents, and other hazardous materials. Bulk gloves are available in various materials, such as latex, nitrile, and vinyl, and can be purchased in large quantities at a lower cost. Choosing the right gloves is crucial, as they should be resistant to the chemicals and substances being handled and should fit properly to avoid contamination.
  • Latex gloves
  • Nitrile gloves
  • Vinyl gloves 
KingFisher automated purification systems
Apex Dx nucleic acid extraction system with mountain background
Scalable services for lab supplies
Bulk magnetic beads and custom manufacturing
Magnetic beads are used in a variety of applications, including DNA and protein purification, cell isolation, and immunoassays. Bulk and OEM magnetic beads can be purchased in large quantities, helping reduce the cost of laboratory operations and enabling consistency and reliability of results. To purchase bulk magnetic beads for extraction and purification of a given analyte, please contact our customer services team or fill out a request for information. 
Magnetic bead-based extraction
Specialty chemical services
Specialty chemical services can provide custom synthesis of chemicals, peptides, and proteins for specific research applications. Bulk specialty chemical services can be purchased at a lower cost, providing researchers with a reliable and cost-effective source of custom chemicals. Specialty lab chemical services include procurement and handling of: 
  • Solvents
  • Buffers and solutions
  • Salts and inorganics
  • Catalysts
  • Acids and bases
  • Pure elements, metals and materials
  • Life science reagents
  • Pharma building blocks
Dynabeads technology OEM partnerships
With 35+ years of experience in designing, developing, and manufacturing paramagnetic beads to internationally recognized quality standards, our Dynabeads OEM team has collaborated with leading biotech, pharma, diagnostic, and healthcare companies. We provide optimized and validated Dynabeads magnetic beads under custom OEM supply arrangements.
Apex Dx nucleic acid extraction system with mountain background
Resources and support
Featured resources for PCR sample preparation
Molecular Cloning Handbook
Invitrogen School of Molecular Biology (ISMB)
Prep for the future with KingFisher instruments—Automated sample preparation for DNA, RNA, protein, or cells
RNA reference guide
Accelerating the future of liquid biopsy: Ultra-low mutation detection solutions from sample prep to data analysis
Dynabeads magnetic beads